Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136543.1 | NP_001130015.1 | 2 | 231 | ||
NM_212551.4 | NP_997716.1 | 2 | 845 | |||
GenBank mRNA | AK298706.1 | - | 2 | 231 | ||
AK298814.1 | - | 2 | 233 | |||
AY037156.1 | - | 2 | 267 | |||
BC031649.1 | - | 2 | 449 | 5172124 | ||
BX647911.1 | - | 2 | 811 |