Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136543.1 | NP_001130015.1 | 1, 2 | 213 | ||
NM_212551.4 | NP_997716.1 | 1, 2 | 827 | |||
GenBank mRNA | AK298706.1 | - | 1, 2 | 213 | ||
AK298814.1 | - | 1, 2 | 215 | |||
AY037156.1 | - | 1, 2 | 249 | |||
BC031649.1 | - | 1, 2 | 431 | 5172124 | ||
BX647911.1 | - | 1, 2 | 793 |