Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOXC1 SIRNA Assays ›
Gene Symbol:
FOXC1
Gene Name:
forkhead box C1
Gene Aliases:
ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3
Chromosome Location:
Chr.6: 1610446 - 1613897 on Build GRCh38
UniGene ID:
Hs.348883
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s5227
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOXC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001453.2
NP_001444.2
1
1549
GenBank mRNA
BC134421.1
-
2
637
Back To Top
More Information
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
ossification
ovarian follicle development
eye development
ureteric bud development
in utero embryonic development
somitogenesis
lymph vessel development
blood vessel remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
brain development
heart development
cell proliferation
germ cell migration
mesenchymal cell development
neural crest cell development
cell migration
negative regulation of angiogenesis
collagen fibril organization
glycosaminoglycan metabolic process
lacrimal gland development
embryonic heart tube development
maintenance of lens transparency
vascular endothelial growth factor signaling pathway
odontogenesis of dentin-containing tooth
camera-type eye development
positive regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
regulation of organ growth
vascular endothelial growth factor receptor signaling pathway
paraxial mesoderm formation
artery morphogenesis
regulation of blood vessel size
ventricular cardiac muscle tissue morphogenesis
cardiac muscle cell proliferation
negative regulation of lymphangiogenesis
positive regulation of hematopoietic progenitor cell differentiation
positive regulation of hematopoietic stem cell differentiation
negative regulation of apoptotic process involved in outflow tract morphogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
DNA binding, bending
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-sv4sw:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0