Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013649.3 | NP_001013671.2 | 4 | 532 | ||
GenBank mRNA | AK300003.1 | - | 4 | 511 | ||
AK311297.1 | - | 3 | 557 | |||
BC111483.1 | - | 4 | 505 | 3877097 | ||
JX960761.1 | - | 4 | 460 |