Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099334.2 | NP_001092804.1 | 6 | 485 | ||
XM_017004074.1 | XP_016859563.1 | 6 | 498 | |||
XM_017004075.1 | XP_016859564.1 | 6 | 433 | |||
XM_017004076.1 | XP_016859565.1 | 6 | 473 | |||
XM_017004077.1 | XP_016859566.1 | 6 | 433 | |||
XM_017004078.1 | XP_016859567.1 | 6 | 433 | |||
GenBank mRNA | BC035737.1 | - | 6 | 485 | 5730487 |