Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR51D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004751.2 | NP_001004751.1 | 1 | 766 | ||
GenBank mRNA | BC140717.1 | - | 1 | 766 | 9021585 |