Search
Search

GPR152| Gene Symbol: GPR152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206997.1 | NP_996880.1 | 1 | 1005 | ||
| GenBank mRNA | AY569571.1 | - | 1 | 1005 | ||
| BC119779.1 | - | 1 | 1005 | 40109407 | ||
| BC122869.1 | - | 1 | 1005 | 40109404 | ||
Set Membership: |
|