Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005467.1 | NP_001005467.1 | 1 | 342 | ||
XM_017017716.1 | XP_016873205.1 | 6 | 1708 | |||
XM_017017717.1 | XP_016873206.1 | 6 | 1011 | |||
XM_017017718.1 | XP_016873207.1 | 4 | 1439 |