Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CBS SIRNA Assays ›
          Gene Symbol:
          CBS CBSL
          Gene Name:
          cystathionine-beta-synthase
          cystathionine-beta-synthase like
          Gene Aliases:
          HIP4
          -
          Chromosome Location:
          Chr.21: 43053190 - 43076868 on Build GRCh38
          Chromosome Location:
          Chr.21: 6444868 - 6468606 on Build GRCh38
          UniGene ID:
          Hs.533013
          Hs.533013
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s528457

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CBS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000071.2 NP_000062.1 17 2153
          NM_001178008.2 NP_001171479.1 17 2160
          NM_001178009.2 NP_001171480.1 18 1946
          NM_001320298.1 NP_001307227.1 18 2154
          NM_001321072.1 NP_001308001.1 14 2219
          XM_011529773.2 XP_011528075.1 16 2321
          XM_011529774.2 XP_011528076.1 17 2107
          XM_011529775.2 XP_011528077.1 15 2279
          XM_011529776.2 XP_011528078.1 16 2065
          XM_011529777.1 XP_011528079.1 18 2138
          XM_011529781.1 XP_011528083.1 19 2203
          XM_011529782.1 XP_011528084.1 18 2601
          XM_011529783.2 XP_011528085.1 15 2233
          XM_017028489.1 XP_016883978.1 19 2598
          XM_017028490.1 XP_016883979.1 18 2553
          XM_017028491.1 XP_016883980.1 18 1882
          XM_017028492.1 XP_016883981.1 19 1947
          XM_017028493.1 XP_016883982.1 15 2432
          GenBank mRNA AK294608.1 - 19 2025
          AL137314.1 - 14 1680
          BC000440.2 - 17 2023 2820694
          BC010242.1 - 17 2077 4154798
          BC011381.1 - 17 2016 3869615
          BC012319.1 - 6 830
          L00972.1 - 14 2231
          L14577.1 - 17 2066
          L19501.1 - 17 2089
          X82166.1 - 17 2055
          Gene Symbol: CBSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321073.1 NP_001308002.1 18 1946
          XM_011546094.1 XP_011544396.1 18 2094
          XM_011546095.2 XP_011544397.1 18 2236
          XM_011546097.2 XP_011544399.1 19 2263
          XM_011546098.1 XP_011544400.1 18 2601
          XM_011546099.1 XP_011544401.1 17 2052
          XM_017028209.1 XP_016883698.1 15 2257
          XM_017028210.1 XP_016883699.1 16 2043
          XM_017028211.1 XP_016883700.1 19 2599
          XM_017028212.1 XP_016883701.1 20 2655
          XM_017028213.1 XP_016883702.1 19 1880
          XM_017028214.1 XP_016883703.1 18 2554
          XM_017028215.1 XP_016883704.1 18 2169
          XM_017028216.1 XP_016883705.1 17 2559
          XM_017028217.1 XP_016883706.1 18 1838
          XM_017028218.1 XP_016883707.1 19 2007
          GenBank mRNA BC007257.1 - 18 1916 3028099

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lyase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cysteine biosynthetic process from serine
          L-serine metabolic process
          L-serine catabolic process
          cellular amino acid biosynthetic process
          cysteine biosynthetic process via cystathionine
          cysteine biosynthetic process
          transsulfuration
          L-cysteine catabolic process
          DNA protection
          homocysteine catabolic process
          homocysteine metabolic process
          oxidation-reduction process
          hydrogen sulfide biosynthetic process
          cystathionine beta-synthase activity
          cysteine synthase activity
          protein binding
          oxygen binding
          enzyme binding
          heme binding
          pyridoxal phosphate binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          nitrite reductase (NO-forming) activity
          carbon monoxide binding
          nitric oxide binding
          modified amino acid binding
          S-adenosyl-L-methionine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-78sq5:80/100.66.76.91:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline