Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEN1 SIRNA Assays ›
          Gene Symbol:
          MEN1
          Gene Name:
          menin 1
          Gene Aliases:
          MEAI, SCG2
          Chromosome Location:
          Chr.11: 64803514 - 64811294 on Build GRCh38
          UniGene ID:
          Hs.423348
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s528732

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000244.3 NP_000235.2 10 2098
          NM_130799.2 NP_570711.1 10 2083
          NM_130800.2 NP_570712.1 10 2061
          NM_130801.2 NP_570713.1 10 2049
          NM_130802.2 NP_570714.1 10 2485
          NM_130803.2 NP_570715.1 10 2492
          NM_130804.2 NP_570716.1 11 2364
          XM_005274001.4 XP_005274058.1 10 2465
          XM_011545040.1 XP_011543342.1 11 2591
          XM_011545041.2 XP_011543343.1 11 2290
          XM_011545042.2 XP_011543344.1 11 2195
          XM_017017765.1 XP_016873254.1 11 2606
          XM_017017766.1 XP_016873255.1 11 2305
          XM_017017767.1 XP_016873256.1 11 2212
          XM_017017768.1 XP_016873257.1 11 2237
          XM_017017769.1 XP_016873258.1 10 2164
          XM_017017770.1 XP_016873259.1 10 2067
          GenBank mRNA BC002544.2 - 10 1936 3138471
          BC002664.2 - 4 992
          U93236.1 - 10 2082
          Y12338.1 - 6 1167

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          mitotic cell cycle
          negative regulation of protein phosphorylation
          osteoblast development
          type B pancreatic cell differentiation
          DNA repair
          transcription, DNA-templated
          cellular response to DNA damage stimulus
          brain development
          negative regulation of cell proliferation
          response to UV
          response to gamma radiation
          negative regulation of cell-substrate adhesion
          positive regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of protein binding
          regulation of activin receptor signaling pathway
          histone lysine methylation
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of osteoblast differentiation
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of JNK cascade
          decidualization
          negative regulation of epithelial cell proliferation
          negative regulation of telomerase activity
          regulation of type B pancreatic cell proliferation
          cellular response to glucose stimulus
          cellular response to peptide hormone stimulus
          response to transforming growth factor beta
          negative regulation of cell cycle G1/S phase transition
          beta-catenin-TCF complex assembly
          four-way junction DNA binding
          Y-form DNA binding
          chromatin binding
          double-stranded DNA binding
          protein binding
          histone-lysine N-methyltransferase activity
          protein binding, bridging
          transcription regulatory region DNA binding
          protein N-terminus binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0