Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGFR2 SIRNA Assays ›
          Gene Symbol:
          FGFR2
          Gene Name:
          fibroblast growth factor receptor 2
          Gene Aliases:
          BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
          Chromosome Location:
          Chr.10: 121478330 - 121598656 on Build GRCh38
          UniGene ID:
          Hs.533683
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529187

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGFR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000141.4 NP_000132.3 11, 12 2202
          NM_001144913.1 NP_001138385.1 10, 11 1708
          NM_001144914.1 NP_001138386.1 8, 9 1369
          NM_001144915.1 NP_001138387.1 10, 11 1607
          NM_001144916.1 NP_001138388.1 8, 9 1651
          NM_001144917.1 NP_001138389.1 9, 10 1854
          NM_001144918.1 NP_001138390.1 9, 10 1851
          NM_001144919.1 NP_001138391.1 10, 11 1938
          NM_001320654.1 NP_001307583.1 10, 11 1586
          NM_001320658.1 NP_001307587.1 11, 12 2196
          NM_022970.3 NP_075259.4 11, 12 2205
          NM_023029.2 NP_075418.1 9, 10 1438
          NR_073009.1 10, 11 2005
          XM_006717708.2 XP_006717771.1 11, 12 2397
          XM_006717710.3 XP_006717773.1 11, 12 2403
          XM_006717711.2 XP_006717774.1 9, 10 1441
          XM_006717712.2 XP_006717775.1 8, 9 1363
          XM_017015920.1 XP_016871409.1 11, 12 2397
          XM_017015921.1 XP_016871410.1 11, 12 2394
          XM_017015922.1 XP_016871411.1 9, 10 1435
          XM_017015923.1 XP_016871412.1 9, 10 1432
          XM_017015924.1 XP_016871413.1 9, 10 2055
          XM_017015925.1 XP_016871414.1 9, 10 2049
          GenBank mRNA AB030073.1 - 11, 12 1976
          AB030074.1 - 11, 12 1976
          AB030075.1 - 11, 12 1976
          AB030076.1 - 11, 12 1976
          AB030077.1 - 11, 12 1976
          AB030078.1 - 11, 12 1976
          AB084153.1 - 8, 9 1219
          AK026508.1 - 10, 11 1912
          AK294026.1 - 11, 12 1607
          AK308621.1 - 8, 9 1651
          BC037338.2 - 10, 11 1586
          BC039243.1 - 9, 10 1851 5277896
          EF057068.1 - 9, 10 1288
          KP192323.1 - 9, 10 1369
          KP192324.1 - 9, 10 1375
          KP192325.1 - 6, 7 1000
          KP192326.1 - 9, 10 1366
          KP192327.1 - 9, 10 1372
          KP192328.1 - 10, 11 1582
          KP192329.1 - 8, 9 1237
          KP192330.1 - 9, 10 1315
          KP192331.1 - 10, 11 1588
          M55614.1 - 10, 11 1570
          M80634.1 - 11, 12 1976
          M87770.1 - 11, 12 1828
          M87771.1 - 9, 10 1694
          M97193.1 - 11, 12 2169
          X52832.1 - 11, 12 1734
          X56191.1 - 10, 11 1582
          Z71929.1 - 10, 11 1567

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          angiogenesis
          ureteric bud development
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of mesenchymal cell proliferation
          outflow tract septum morphogenesis
          membranous septum morphogenesis
          apoptotic process
          cell-cell signaling
          axonogenesis
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          regulation of smoothened signaling pathway
          post-embryonic development
          embryonic pattern specification
          organ morphogenesis
          regulation of cell fate commitment
          positive regulation of phospholipase activity
          regulation of phosphatidylinositol 3-kinase signaling
          morphogenesis of embryonic epithelium
          peptidyl-tyrosine phosphorylation
          orbitofrontal cortex development
          ventricular zone neuroblast division
          pyramidal neuron development
          gland morphogenesis
          positive regulation of Wnt signaling pathway
          bone mineralization
          lung development
          epithelial cell differentiation
          midbrain development
          otic vesicle formation
          hair follicle morphogenesis
          lacrimal gland development
          regulation of osteoblast proliferation
          multicellular organism growth
          organ growth
          fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
          fibroblast growth factor receptor signaling pathway involved in hemopoiesis
          fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          phosphatidylinositol-3-phosphate biosynthetic process
          regulation of multicellular organism growth
          regulation of fibroblast growth factor receptor signaling pathway
          inner ear morphogenesis
          odontogenesis
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          cell fate commitment
          regulation of osteoblast differentiation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          lung alveolus development
          mesodermal cell differentiation
          embryonic digestive tract morphogenesis
          embryonic organ morphogenesis
          digestive tract development
          embryonic organ development
          reproductive structure development
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          epidermis morphogenesis
          branching morphogenesis of a nerve
          mesenchymal cell differentiation
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          regulation of smooth muscle cell differentiation
          positive regulation of cell division
          ventricular cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          limb bud formation
          bone development
          bone morphogenesis
          branching involved in prostate gland morphogenesis
          branching involved in salivary gland morphogenesis
          bud elongation involved in lung branching
          lung lobe morphogenesis
          lung-associated mesenchyme development
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          prostate gland morphogenesis
          prostate epithelial cord elongation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
          fibroblast growth factor receptor signaling pathway involved in mammary gland specification
          lateral sprouting from an epithelium
          mammary gland bud formation
          epithelial cell proliferation involved in salivary gland morphogenesis
          branch elongation involved in salivary gland morphogenesis
          branching involved in labyrinthine layer morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          regulation of morphogenesis of a branching structure
          mesenchymal cell differentiation involved in lung development
          mesenchymal cell proliferation involved in lung development
          regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of canonical Wnt signaling pathway
          protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          fibroblast growth factor binding
          protein homodimerization activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0