Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MID1 SIRNA Assays ›
Gene Symbol:
MID1
Gene Name:
midline 1
Gene Aliases:
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location:
Chr.X: 10445310 - 11111264 on Build GRCh38
UniGene ID:
Hs.27695
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529208
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MID1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000381.3
NP_000372.1
3
1092
NM_001098624.2
NP_001092094.1
3
1022
NM_001193277.1
NP_001180206.1
3
881
NM_001193278.1
NP_001180207.1
2
747
NM_001193279.1
NP_001180208.1
2
747
NM_001193280.1
NP_001180209.1
2
633
NM_001193281.1
NP_001180210.1
2
633
NM_033289.1
NP_150631.1
3
978
NM_033290.3
NP_150632.1
3
884
XM_005274536.1
XP_005274593.1
3
904
XM_005274537.1
XP_005274594.1
3
881
XM_006724492.3
XP_006724555.1
3
1503
XM_011545525.2
XP_011543827.1
5
1205
XM_011545526.2
XP_011543828.1
5
1240
XM_011545527.2
XP_011543829.1
3
1150
XM_017029534.1
XP_016885023.1
3
1070
XM_017029535.1
XP_016885024.1
5
1091
XM_017029536.1
XP_016885025.1
5
1320
XM_017029537.1
XP_016885026.1
5
1091
XM_017029538.1
XP_016885027.1
3
790
XM_017029539.1
XP_016885028.1
3
952
XM_017029540.1
XP_016885029.1
5
1205
XM_017029541.1
XP_016885030.1
5
1205
XM_017029542.1
XP_016885031.1
5
1205
GenBank mRNA
AF035360.1
-
3
778
AF041206.1
-
3
972
AF041208.1
-
3
1092
AF041210.1
-
2
581
AF230976.1
-
3
877
AF230977.1
-
3
877
AF269101.1
-
4
1000
AK025187.1
-
2
63
AK226104.1
-
3
796
AK291215.1
-
3
974
AK297205.1
-
4
888
AK299061.1
-
3
917
AK315095.1
-
3
923
AY539962.1
-
2
74
AY539963.1
-
3
235
AY539964.1
-
2
74
AY539965.1
-
2
74
AY539976.1
-
2
160
AY539988.1
-
2
577
AY539989.1
-
2
306
AY539990.1
-
3
558
AY539994.1
-
2
691
AY539995.1
-
2
306
AY539996.1
-
2
192
AY540001.1
-
2
306
AY540002.1
-
2
192
AY540014.1
-
2
192
AY540015.1
-
2
306
AY540016.1
-
2
306
AY540020.1
-
2
306
AY540022.1
-
2
691
AY540023.1
-
2
577
BC053626.1
-
3
949
6163881
Y13667.1
-
3
877
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
pattern specification process
positive regulation of stress-activated MAPK cascade
protein localization to microtubule
interferon-gamma-mediated signaling pathway
protein binding
microtubule binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline