Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAX6 SIRNA Assays ›
          Gene Symbol:
          PAX6
          Gene Name:
          paired box 6
          Gene Aliases:
          AN, AN2, D11S812E, FVH1, MGDA, WAGR
          Chromosome Location:
          Chr.11: 31784792 - 31817961 on Build GRCh38
          UniGene ID:
          Hs.270303
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529238

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PAX6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000280.4 NP_000271.1 10 1405
          NM_001127612.1 NP_001121084.1 10 1319
          NM_001258462.1 NP_001245391.1 11 1361
          NM_001258463.1 NP_001245392.1 11 1299
          NM_001258464.1 NP_001245393.1 10 1307
          NM_001258465.1 NP_001245394.1 9 1293
          NM_001310158.1 NP_001297087.1 11 1402
          NM_001310159.1 NP_001297088.1 8 978
          NM_001310160.1 NP_001297089.1 7 2616
          NM_001310161.1 NP_001297090.1 8 1168
          NM_001604.5 NP_001595.2 11 1349
          GenBank mRNA AB209177.1 - 8 5757
          AB593092.1 - 11 1300
          AB593093.1 - 11 1299
          AB593094.1 - 11 1402
          AK074881.1 - 9 1740
          AK094172.1 - 5 2043
          AK094249.1 - 7 2426
          AK314470.1 - 10 1297
          AY047583.1 - 8 980
          AY707088.1 - 6 850
          BC011953.1 - 10 1191 3880468
          BI816814.1 - 1 73
          BX640762.1 - 11 1328
          GQ141695.1 - 7 898
          M77844.1 - 9 1212
          M93650.1 - 10 1267

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          establishment of mitotic spindle orientation
          blood vessel development
          eye development
          cell fate determination
          neuron migration
          negative regulation of protein phosphorylation
          positive regulation of neuroblast proliferation
          lens development in camera-type eye
          type B pancreatic cell differentiation
          pancreatic A cell development
          transcription from RNA polymerase II promoter
          smoothened signaling pathway
          axon guidance
          central nervous system development
          salivary gland morphogenesis
          visual perception
          response to wounding
          regulation of asymmetric cell division
          organ morphogenesis
          dorsal/ventral axis specification
          positive regulation of gene expression
          protein ubiquitination
          oligodendrocyte cell fate specification
          cerebral cortex regionalization
          forebrain anterior/posterior pattern specification
          forebrain dorsal/ventral pattern formation
          commitment of neuronal cell to specific neuron type in forebrain
          forebrain-midbrain boundary formation
          regulation of transcription from RNA polymerase II promoter involved in spinal cord motor neuron fate specification
          regulation of transcription from RNA polymerase II promoter involved in ventral spinal cord interneuron specification
          regulation of transcription from RNA polymerase II promoter involved in somatic motor neuron fate commitment
          pituitary gland development
          signal transduction involved in regulation of gene expression
          keratinocyte differentiation
          regulation of cell migration
          positive regulation of epithelial cell differentiation
          lacrimal gland development
          protein localization to organelle
          eye photoreceptor cell development
          glucose homeostasis
          negative regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of timing of cell differentiation
          embryonic camera-type eye morphogenesis
          neuron fate commitment
          astrocyte differentiation
          negative regulation of epithelial cell proliferation
          negative regulation of neurogenesis
          retina development in camera-type eye
          iris morphogenesis
          cornea development in camera-type eye
          negative regulation of neural precursor cell proliferation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          ubiquitin-protein transferase activity
          protein binding
          transcription factor binding
          protein kinase binding
          ubiquitin protein ligase binding
          histone acetyltransferase binding
          co-SMAD binding
          R-SMAD binding
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0