Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCOF1 SIRNA Assays ›
          Gene Symbol:
          TCOF1
          Gene Name:
          treacle ribosome biogenesis factor 1
          Gene Aliases:
          MFD1, TCS, TCS1, treacle
          Chromosome Location:
          Chr.5: 150357639 - 150400308 on Build GRCh38
          UniGene ID:
          Hs.519672
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529266

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCOF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000356.3 NP_000347.2 8 1126
          NM_001008657.2 NP_001008657.1 9 1357
          NM_001135243.1 NP_001128715.1 9 1357
          NM_001135244.1 NP_001128716.1 9 1357
          NM_001135245.1 NP_001128717.1 8 1126
          NM_001195141.1 NP_001182070.1 9 1357
          XM_005268502.3 XP_005268559.1 9 1294
          XM_005268503.3 XP_005268560.1 9 1294
          XM_005268504.3 XP_005268561.1 9 1294
          XM_005268505.3 XP_005268562.1 9 1294
          XM_005268506.3 XP_005268563.1 9 1294
          XM_005268507.3 XP_005268564.1 8 1063
          XM_005268508.3 XP_005268565.1 9 1294
          XM_005268509.3 XP_005268566.1 9 1294
          XM_011537678.2 XP_011535980.1 9 1294
          XM_017009792.1 XP_016865281.1 9 1294
          XM_017009793.1 XP_016865282.1 9 1294
          XM_017009794.1 XP_016865283.1 9 1294
          XM_017009795.1 XP_016865284.1 8 1063
          GenBank mRNA AB209317.1 - 1 49
          AK096467.1 - 5 863
          AK225284.1 - 8 1051
          AK299167.1 - 2 314
          AK303611.1 - 9 1282
          AK307586.1 - 9 1282
          AY460334.1 - 9 1249
          BC011764.2 - 9 1278 3616898
          BC014559.2 - 9 1278 4025673
          BC016144.1 - 9 1277
          BC027252.1 - 8 974
          BC033093.1 - 9 1278 4577077
          BX537490.1 - 5 2014
          U40847.1 - 8 1111
          U76366.1 - 8 1093

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          transport
          transporter activity
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bz4hs:80/100.66.77.47:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0