Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP53 SIRNA Assays ›
Gene Symbol:
TP53
Gene Name:
tumor protein p53
Gene Aliases:
BCC7, LFS1, P53, TRP53
Chromosome Location:
Chr.17: 7668402 - 7687550 on Build GRCh38
UniGene ID:
Hs.437460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529268
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP53
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000546.5
NP_000537.3
6
835
NM_001126112.2
NP_001119584.1
6
832
NM_001126113.2
NP_001119585.1
6
835
NM_001126114.2
NP_001119586.1
6
835
NM_001126115.1
NP_001119587.1
2
515
NM_001126116.1
NP_001119588.1
2
515
NM_001126117.1
NP_001119589.1
2
515
NM_001126118.1
NP_001119590.1
5
952
NM_001276695.1
NP_001263624.1
6
835
NM_001276696.1
NP_001263625.1
6
835
NM_001276697.1
NP_001263626.1
2
515
NM_001276698.1
NP_001263627.1
2
515
NM_001276699.1
NP_001263628.1
2
515
NM_001276760.1
NP_001263689.1
6
835
NM_001276761.1
NP_001263690.1
6
832
GenBank mRNA
AB082923.1
-
6
696
AF307851.1
-
6
768
AK225838.1
-
6
773
AK297462.1
-
7
700
AK297927.1
-
7
745
AK303277.1
-
5
891
AK312568.1
-
6
777
AM076970.1
-
5
600
AM076971.1
-
5
633
AM076972.1
-
5
633
BC003596.1
-
6
753
3544714
BT019622.1
-
5
633
DQ186648.1
-
6
765
DQ186649.1
-
6
765
DQ186650.1
-
2
511
DQ186651.1
-
2
511
DQ186652.1
-
2
511
DQ191317.1
-
6
822
DQ263704.1
-
5
633
DQ286964.1
-
6
767
DQ648884.1
-
5
433
DQ648885.1
-
3
309
EF101867.1
-
5
600
EF101868.1
-
5
633
EF101869.1
-
5
633
FJ207420.1
-
6
768
JN214348.1
-
5
678
JN601513.1
-
5
633
JN601514.1
-
5
633
JN601515.1
-
5
633
JN601516.1
-
6
607
JN900492.1
-
5
941
JQ026207.1
-
5
623
JQ026208.1
-
5
628
JQ026209.1
-
5
633
JQ026210.1
-
5
633
JQ694049.1
-
5
656
JQ694050.1
-
5
656
JQ694051.1
-
5
656
KC820708.1
-
2
128
KC820709.1
-
2
128
KC820710.1
-
2
128
KC820711.1
-
2
128
KC820712.1
-
2
128
KC820713.1
-
2
128
KC820714.1
-
2
128
KC820715.1
-
2
128
KC820716.1
-
2
128
KC820717.1
-
2
128
KC820718.1
-
2
128
KC820719.1
-
2
128
KC820720.1
-
2
128
KC820721.1
-
2
128
KC820722.1
-
2
128
KC820723.1
-
2
128
KC820724.1
-
2
128
KC820725.1
-
2
128
KC820726.1
-
2
128
KC820727.1
-
2
128
KC820728.1
-
2
128
KC820729.1
-
2
128
KC820730.1
-
2
128
KC820731.1
-
2
128
KC820732.1
-
2
128
KC820733.1
-
2
128
KC820734.1
-
2
128
KC820735.1
-
2
128
KC820736.1
-
2
128
KC820737.1
-
2
128
KC820738.1
-
2
128
KC820739.1
-
2
128
KC820740.1
-
2
128
KC820741.1
-
2
128
KC820742.1
-
2
128
KC820743.1
-
2
128
KC820744.1
-
2
128
KC820745.1
-
2
128
KC820746.1
-
2
128
KC820747.1
-
2
128
KC820748.1
-
2
128
KC820749.1
-
2
128
KC820750.1
-
2
128
KC820751.1
-
2
128
KC820752.1
-
2
128
KC820753.1
-
2
128
KC820754.1
-
2
128
KC820755.1
-
2
128
KC820756.1
-
2
128
KC820757.1
-
2
128
KC820758.1
-
2
128
KC820760.1
-
2
128
KC820761.1
-
2
128
KC820762.1
-
2
128
KC820763.1
-
2
128
KC820764.1
-
2
128
KC820765.1
-
2
128
KC820766.1
-
2
128
KC820767.1
-
2
128
KC820768.1
-
2
128
KC820769.1
-
2
128
KC820770.1
-
2
128
KC820771.1
-
2
128
KC820772.1
-
2
128
KC820773.1
-
2
128
KC820774.1
-
2
128
KC820775.1
-
2
128
KC820776.1
-
2
128
KC820777.1
-
2
128
KC820778.1
-
2
128
KC820779.1
-
2
128
KC820780.1
-
2
128
KC820781.1
-
2
128
KC820782.1
-
2
128
KC820783.1
-
2
128
KC820784.1
-
2
128
KC820785.1
-
2
128
KC820786.1
-
2
128
M14694.1
-
6
758
M14695.1
-
6
754
X01405.1
-
3
334
X02469.1
-
6
768
X60010.1
-
5
632
X60011.1
-
5
633
X60012.1
-
5
633
X60013.1
-
5
633
X60014.1
-
5
633
X60015.1
-
5
633
X60016.1
-
5
633
X60017.1
-
5
633
X60018.1
-
5
633
X60020.1
-
5
633
Back To Top
More Information
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA strand renaturation
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
ER overload response
cell cycle arrest
Ras protein signal transduction
multicellular organism development
cell aging
protein localization
cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
response to X-ray
response to gamma radiation
positive regulation of gene expression
viral process
protein sumoylation
cell differentiation
negative regulation of cell growth
DNA damage response, signal transduction by p53 class mediator
positive regulation of histone deacetylation
chromatin assembly
mitotic G1 DNA damage checkpoint
positive regulation of protein oligomerization
cellular response to UV
cellular response to drug
cellular response to glucose starvation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
positive regulation of protein export from nucleus
regulation of mitochondrial membrane permeability
negative regulation of fibroblast proliferation
circadian behavior
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of helicase activity
protein tetramerization
negative regulation of telomerase activity
positive regulation of thymocyte apoptotic process
positive regulation of cell cycle arrest
cellular response to hypoxia
cellular response to ionizing radiation
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
replicative senescence
oxidative stress-induced premature senescence
intrinsic apoptotic signaling pathway
oligodendrocyte apoptotic process
positive regulation of execution phase of apoptosis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
regulation of cell cycle G2/M phase transition
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of reactive oxygen species metabolic process
positive regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protease binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
copper ion binding
protein binding
ATP binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
protein phosphatase binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
identical protein binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
chaperone binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0