Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP53 SIRNA Assays ›
Gene Symbol:
TP53
Gene Name:
tumor protein p53
Gene Aliases:
BCC7, LFS1, P53, TRP53
Chromosome Location:
Chr.17: 7668402 - 7687550 on Build GRCh38
UniGene ID:
Hs.437460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529270
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP53
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000546.5
NP_000537.3
7
886
NM_001126112.2
NP_001119584.1
7
883
NM_001126113.2
NP_001119585.1
7
886
NM_001126114.2
NP_001119586.1
7
886
NM_001126115.1
NP_001119587.1
3
566
NM_001126116.1
NP_001119588.1
3
566
NM_001126117.1
NP_001119589.1
3
566
NM_001126118.1
NP_001119590.1
6
1003
NM_001276695.1
NP_001263624.1
7
886
NM_001276696.1
NP_001263625.1
7
886
NM_001276697.1
NP_001263626.1
3
566
NM_001276698.1
NP_001263627.1
3
566
NM_001276699.1
NP_001263628.1
3
566
NM_001276760.1
NP_001263689.1
7
886
NM_001276761.1
NP_001263690.1
7
883
GenBank mRNA
AB082923.1
-
7
747
AF307851.1
-
7
819
AK225838.1
-
7
824
AK297462.1
-
8
751
AK297927.1
-
8
796
AK312568.1
-
7
828
AM076970.1
-
6
651
AM076971.1
-
6
684
AM076972.1
-
6
684
AY429684.1
-
6
684
BC003596.1
-
7
804
3544714
BT019622.1
-
6
684
DQ186648.1
-
7
816
DQ186649.1
-
7
816
DQ186650.1
-
3
562
DQ186651.1
-
3
562
DQ186652.1
-
3
562
DQ191317.1
-
7
873
DQ263704.1
-
6
684
DQ286964.1
-
7
818
DQ485152.1
-
2
79
DQ648884.1
-
6
484
DQ648885.1
-
4
360
EF101867.1
-
6
651
EF101868.1
-
6
684
EF101869.1
-
6
684
FJ207420.1
-
7
819
JN214348.1
-
6
729
JN601513.1
-
6
684
JN601514.1
-
6
684
JN601515.1
-
6
684
JN601516.1
-
7
658
JN900492.1
-
6
992
JQ026207.1
-
6
674
JQ026208.1
-
6
679
JQ026209.1
-
6
689
JQ026210.1
-
6
684
JQ694049.1
-
6
707
JQ694050.1
-
6
707
JQ694051.1
-
6
707
KC820708.1
-
3
179
KC820709.1
-
3
179
KC820710.1
-
3
179
KC820711.1
-
3
179
KC820712.1
-
3
179
KC820713.1
-
3
179
KC820714.1
-
3
179
KC820715.1
-
3
179
KC820716.1
-
3
179
KC820717.1
-
3
179
KC820718.1
-
3
179
KC820719.1
-
3
179
KC820720.1
-
3
179
KC820721.1
-
3
179
KC820722.1
-
3
179
KC820723.1
-
3
179
KC820724.1
-
3
179
KC820725.1
-
3
179
KC820726.1
-
3
179
KC820727.1
-
3
179
KC820728.1
-
3
179
KC820729.1
-
3
179
KC820730.1
-
3
179
KC820731.1
-
3
179
KC820732.1
-
3
179
KC820733.1
-
3
179
KC820734.1
-
3
179
KC820735.1
-
3
179
KC820736.1
-
3
179
KC820737.1
-
3
179
KC820738.1
-
3
179
KC820739.1
-
3
179
KC820740.1
-
3
179
KC820741.1
-
3
179
KC820742.1
-
3
179
KC820743.1
-
3
179
KC820744.1
-
3
179
KC820745.1
-
3
179
KC820746.1
-
3
179
KC820747.1
-
3
179
KC820748.1
-
3
179
KC820749.1
-
3
179
KC820750.1
-
3
179
KC820751.1
-
3
179
KC820752.1
-
3
179
KC820753.1
-
3
179
KC820754.1
-
3
179
KC820755.1
-
3
179
KC820756.1
-
3
179
KC820757.1
-
3
179
KC820758.1
-
3
179
KC820759.1
-
2
68
KC820760.1
-
3
179
KC820761.1
-
3
179
KC820762.1
-
3
179
KC820763.1
-
3
179
KC820764.1
-
3
179
KC820765.1
-
3
179
KC820766.1
-
3
179
KC820767.1
-
3
179
KC820768.1
-
3
179
KC820769.1
-
3
179
KC820770.1
-
3
179
KC820771.1
-
3
179
KC820772.1
-
3
179
KC820773.1
-
3
179
KC820774.1
-
3
179
KC820775.1
-
3
179
KC820776.1
-
3
179
KC820777.1
-
3
179
KC820778.1
-
3
179
KC820779.1
-
3
179
KC820780.1
-
3
179
KC820781.1
-
3
179
KC820782.1
-
3
179
KC820783.1
-
3
179
KC820784.1
-
3
179
KC820785.1
-
3
179
KC820786.1
-
3
179
M14694.1
-
7
809
M14695.1
-
7
805
X01405.1
-
4
385
X02469.1
-
7
819
X60010.1
-
6
683
X60011.1
-
6
684
X60012.1
-
6
684
X60013.1
-
6
684
X60014.1
-
6
684
X60015.1
-
6
684
X60016.1
-
6
684
X60017.1
-
6
684
X60018.1
-
6
684
X60019.1
-
6
684
X60020.1
-
6
684
Back To Top
More Information
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA strand renaturation
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
ER overload response
cell cycle arrest
Ras protein signal transduction
multicellular organism development
cell aging
protein localization
cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
response to X-ray
response to gamma radiation
positive regulation of gene expression
viral process
protein sumoylation
cell differentiation
negative regulation of cell growth
DNA damage response, signal transduction by p53 class mediator
positive regulation of histone deacetylation
chromatin assembly
mitotic G1 DNA damage checkpoint
positive regulation of protein oligomerization
cellular response to UV
cellular response to drug
cellular response to glucose starvation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
positive regulation of protein export from nucleus
regulation of mitochondrial membrane permeability
negative regulation of fibroblast proliferation
circadian behavior
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of helicase activity
protein tetramerization
negative regulation of telomerase activity
positive regulation of thymocyte apoptotic process
positive regulation of cell cycle arrest
cellular response to hypoxia
cellular response to ionizing radiation
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
replicative senescence
oxidative stress-induced premature senescence
intrinsic apoptotic signaling pathway
oligodendrocyte apoptotic process
positive regulation of execution phase of apoptosis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
regulation of cell cycle G2/M phase transition
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of reactive oxygen species metabolic process
positive regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protease binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
copper ion binding
protein binding
ATP binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
protein phosphatase binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
identical protein binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
chaperone binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0