Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BEST1 SIRNA Assays ›
          Gene Symbol:
          BEST1
          Gene Name:
          bestrophin 1
          Gene Aliases:
          ARB, BEST, BMD, RP50, TU15B, VMD2
          Chromosome Location:
          Chr.11: 61946722 - 61964465 on Build GRCh38
          UniGene ID:
          Hs.524910
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529275

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BEST1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001139443.1 NP_001132915.1 8 1413
          NM_001300786.1 NP_001287715.1 7 1332
          NM_001300787.1 NP_001287716.1 8 1413
          NM_004183.3 NP_004174.1 9 1601
          NR_134580.1 9 1804
          XM_005274210.3 XP_005274267.1 9 1711
          XM_005274215.3 XP_005274272.1 7 1366
          XM_005274216.3 XP_005274273.1 8 1726
          XM_011545229.2 XP_011543531.1 9 1151
          XM_011545230.2 XP_011543532.1 8 954
          XM_011545233.2 XP_011543535.1 3 848
          XM_017018228.1 XP_016873717.1 7 1366
          XM_017018229.1 XP_016873718.1 9 1914
          XM_017018230.1 XP_016873719.1 7 1569
          GenBank mRNA AB209451.1 - 6 2642
          AF057169.1 - 9 1125
          AF057170.1 - 9 1328
          AF073501.1 - 9 1109
          AK289681.1 - 9 1332
          AK293701.1 - 8 1067
          AK295450.1 - 8 1147
          AK295998.1 - 8 947
          AK301392.1 - 7 930
          AY515704.1 - 8 1021
          BC015220.2 - 7 1332 3877806
          BC041664.1 - 8 927 5194649
          JQ954696.1 - 8 1087
          KU178390.1 - 6 760
          KU178391.1 - 7 841
          KU178392.1 - 6 688

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transport
          visual perception
          bicarbonate transport
          transepithelial chloride transport
          ion transmembrane transport
          detection of light stimulus involved in visual perception
          regulation of calcium ion transport
          chloride transmembrane transport
          intracellular calcium activated chloride channel activity
          chloride channel activity
          bicarbonate transmembrane transporter activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0