Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDH23 SIRNA Assays ›
Gene Symbol:
CDH23
Gene Name:
cadherin-related 23
Gene Aliases:
CDHR23, USH1D
Chromosome Location:
Chr.10: 71396934 - 71815947 on Build GRCh38
UniGene ID:
Hs.656032
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529350
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDH23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171930.1
NP_001165401.1
11
1405
NM_001171931.1
NP_001165402.1
11
1405
NM_001171932.1
NP_001165403.1
11
1405
NM_022124.5
NP_071407.4
11
1405
NM_052836.3
NP_443068.1
11
1405
XM_006717940.3
XP_006718003.1
11
1399
XM_006717942.3
XP_006718005.1
11
1399
XM_011540039.2
XP_011538341.1
11
1399
XM_011540042.2
XP_011538344.1
11
1399
XM_011540043.2
XP_011538345.1
11
1399
XM_011540044.2
XP_011538346.1
11
1040
XM_011540045.2
XP_011538347.1
11
1399
XM_011540046.2
XP_011538348.1
6
834
XM_011540048.2
XP_011538350.1
11
1399
XM_011540049.2
XP_011538351.1
11
1399
XM_011540051.2
XP_011538353.1
11
1399
XM_017016499.1
XP_016871988.1
11
1405
XM_017016500.1
XP_016871989.1
11
1399
GenBank mRNA
AF312024.1
-
10
1015
AK074160.1
-
4
387
AY010111.2
-
10
1015
AY358617.1
-
11
1055
BC011570.1
-
11
1410
BC032581.1
-
11
1411
BC065284.1
-
11
1382
BC108254.1
-
9
1173
BC136976.1
-
11
1275
BC136977.1
-
11
1274
BC139903.1
-
11
1397
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
homophilic cell adhesion via plasma membrane adhesion molecules
visual perception
sensory perception of sound
locomotory behavior
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
photoreceptor cell maintenance
response to stimulus
sensory perception of light stimulus
equilibrioception
regulation of cytosolic calcium ion concentration
inner ear receptor stereocilium organization
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0