Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000153.3 | NP_000144.2 | 13 | 1582 | ||
NM_001201401.1 | NP_001188330.1 | 12 | 1513 | |||
NM_001201402.1 | NP_001188331.1 | 13 | 1563 | |||
XM_011536618.2 | XP_011534920.1 | 13 | 1864 | |||
GenBank mRNA | AK294040.1 | - | 14 | 1427 | ||
AK296530.1 | - | 12 | 1404 | |||
AK302519.1 | - | 8 | 998 | |||
AK302683.1 | - | 13 | 1736 | |||
AK302956.1 | - | 13 | 1402 | |||
BC036518.2 | - | 13 | 1434 | 5267705 | ||
D25283.1 | - | 13 | 1465 | |||
L23116.1 | - | 13 | 1474 |