Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LMNA SIRNA Assays ›
          Gene Symbol:
          LMNA
          Gene Name:
          lamin A/C
          Gene Aliases:
          CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, FPLD2, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, LMNL1, PRO1
          Chromosome Location:
          Chr.1: 156082546 - 156140089 on Build GRCh38
          UniGene ID:
          Hs.594444
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s530949

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LMNA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257374.2 NP_001244303.1 4 523
          NM_001282624.1 NP_001269553.1 5 703
          NM_001282625.1 NP_001269554.1 7 1445
          NM_001282626.1 NP_001269555.1 4 1021
          NM_005572.3 NP_005563.1 4 1021
          NM_170707.3 NP_733821.1 4 1021
          NM_170708.3 NP_733822.1 4 1021
          XM_011509533.1 XP_011507835.1 4 518
          XM_011509534.1 XP_011507836.1 3 416
          GenBank mRNA AK056143.1 - 7 1413
          AK056191.1 - 5 596
          AK057997.1 - 4 870
          AK097801.1 - 5 631
          AK098128.1 - 1 117
          AK122732.1 - 5 907
          AK130179.1 - 4 928
          AK295390.1 - 4 470
          AY357727.1 - 4 969
          AY528714.1 - 4 890
          AY847595.1 - 4 772
          AY847596.1 - 4 772
          AY847597.1 - 4 772
          BC000511.2 - 4 952 2822703
          BC003162.1 - 4 937 2958052
          BC014507.1 - 4 969 4863480
          BC033088.1 - 4 965 3623265
          DC400775.1 - 4 480
          M13451.1 - 4 906
          M13452.1 - 3 325
          X03444.1 - 4 982
          X03445.1 - 4 982

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic nuclear envelope disassembly
          mitotic nuclear envelope reassembly
          spermatogenesis
          response to mechanical stimulus
          regulation of cell migration
          establishment or maintenance of microtubule cytoskeleton polarity
          protein localization to nucleus
          sterol regulatory element binding protein import into nucleus
          IRE1-mediated unfolded protein response
          positive regulation of osteoblast differentiation
          ventricular cardiac muscle cell development
          cellular response to hypoxia
          negative regulation of release of cytochrome c from mitochondria
          positive regulation of cell aging
          regulation of protein localization to nucleus
          negative regulation of adipose tissue development
          negative regulation of extrinsic apoptotic signaling pathway
          structural molecule activity
          protein binding
          protein phosphatase 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0