Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGFR1 SIRNA Assays ›
          Gene Symbol:
          FGFR1
          Gene Name:
          fibroblast growth factor receptor 1
          Gene Aliases:
          BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
          Chromosome Location:
          Chr.8: 38411138 - 38468834 on Build GRCh38
          UniGene ID:
          Hs.264887
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531162

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGFR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001174063.1 NP_001167534.1 12, 13 2604
          NM_001174064.1 NP_001167535.1 13, 14 2672
          NM_001174065.1 NP_001167536.1 12, 13 1924
          NM_001174066.1 NP_001167537.1 11, 12 1663
          NM_001174067.1 NP_001167538.1 13, 14 2084
          NM_015850.3 NP_056934.2 12, 13 2604
          NM_023105.2 NP_075593.1 11, 12 2343
          NM_023106.2 NP_075594.1 11, 12 2337
          NM_023110.2 NP_075598.2 12, 13 2610
          XM_006716303.2 XP_006716366.1 12, 13 2411
          XM_006716304.1 XP_006716367.1 12, 13 1927
          XM_006716306.2 XP_006716369.1 12, 13 2405
          XM_006716307.1 XP_006716370.1 12, 13 1921
          XM_006716309.3 XP_006716372.1 12, 13 2381
          XM_006716310.2 XP_006716373.1 11, 12 2144
          XM_006716311.1 XP_006716374.1 11, 12 1542
          XM_006716312.1 XP_006716375.1 11, 12 1612
          XM_006716313.2 XP_006716376.1 11, 12 2138
          XM_006716314.1 XP_006716377.1 11, 12 1616
          XM_011544443.2 XP_011542745.1 12, 13 1916
          XM_011544444.1 XP_011542746.1 12, 13 1905
          XM_011544445.2 XP_011542747.1 12, 13 1918
          XM_011544446.2 XP_011542748.1 12, 13 1911
          XM_011544447.2 XP_011542749.1 12, 13 1905
          XM_011544448.1 XP_011542750.1 11, 12 1515
          XM_011544449.1 XP_011542751.1 11, 12 1509
          XM_011544450.2 XP_011542752.1 11, 12 1509
          XM_011544451.1 XP_011542753.1 10, 11 1378
          XM_017013219.1 XP_016868708.1 12, 13 1906
          XM_017013220.1 XP_016868709.1 12, 13 1899
          XM_017013221.1 XP_016868710.1 13, 14 2120
          XM_017013222.1 XP_016868711.1 12, 13 2405
          XM_017013223.1 XP_016868712.1 12, 13 1915
          XM_017013224.1 XP_016868713.1 12, 13 2399
          XM_017013225.1 XP_016868714.1 12, 13 1882
          XM_017013226.1 XP_016868715.1 11, 12 1509
          XM_017013227.1 XP_016868716.1 11, 12 1503
          XM_017013228.1 XP_016868717.1 11, 12 2132
          XM_017013229.1 XP_016868718.1 6, 7 2091
          XM_017013230.1 XP_016868719.1 10, 11 2697
          GenBank mRNA AB208919.1 - 13, 14 2459
          AK222718.1 - 12, 13 2403
          AK291754.1 - 12, 13 1803
          AK292470.1 - 11, 12 1663
          AK309947.1 - 13, 14 1963
          BC015035.1 - 13, 14 2332 3911101
          BC018128.1 - 12, 13 2368 3896359
          BC091494.1 - 10, 11 1474 30387634
          BX649167.1 - 8, 9 3952
          FJ809917.1 - 12, 13 2619
          KU178066.1 - 11, 12 1662
          KU178067.1 - 10, 11 1405
          KU178068.1 - 10, 11 1399
          KU178069.1 - 10, 11 1395
          KU178070.1 - 10, 11 1389
          M34185.1 - 11, 12 1656
          M34186.1 - 11, 12 1921
          M34641.1 - 11, 12 1671
          M37722.1 - 11, 12 1623
          M60485.1 - 13, 14 2333
          M63887.1 - 11, 12 1668
          M63888.1 - 11, 12 1668
          M63889.1 - 11, 12 1545
          X51803.1 - 12, 13 1785
          X52833.1 - 11, 12 1755
          X57119.1 - 11, 12 1563
          X57120.1 - 12, 13 1824
          X57121.1 - 12, 13 1830
          X57122.1 - 11, 12 1557
          X66945.1 - 12, 13 2394
          Y00665.1 - 8, 9 1068

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          skeletal system development
          angiogenesis
          ureteric bud development
          in utero embryonic development
          organ induction
          neuron migration
          positive regulation of mesenchymal cell proliferation
          chondrocyte differentiation
          transcription, DNA-templated
          protein phosphorylation
          sensory perception of sound
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          positive regulation of phospholipase C activity
          positive regulation of neuron projection development
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          peptidyl-tyrosine phosphorylation
          stem cell population maintenance
          motogenic signaling involved in postnatal olfactory bulb interneuron migration
          ventricular zone neuroblast division
          embryonic limb morphogenesis
          midbrain development
          neuron projection development
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          phosphatidylinositol-3-phosphate biosynthetic process
          inner ear morphogenesis
          outer ear morphogenesis
          middle ear morphogenesis
          chordate embryonic development
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          regulation of cell differentiation
          positive regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of cell cycle
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          paraxial mesoderm development
          regulation of lateral mesodermal cell fate specification
          cell maturation
          skeletal system morphogenesis
          mesenchymal cell differentiation
          regulation of sensory perception of pain
          positive regulation of cardiac muscle cell proliferation
          auditory receptor cell development
          branching involved in salivary gland morphogenesis
          lung-associated mesenchyme development
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          regulation of stem cell proliferation
          positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
          positive regulation of endothelial cell chemotaxis to fibroblast growth factor
          regulation of extrinsic apoptotic signaling pathway in absence of ligand
          glycoprotein binding
          protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          fibroblast growth factor binding
          protein complex binding
          identical protein binding
          protein homodimerization activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0