Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HINT1 SIRNA Assays ›
          Gene Symbol:
          HINT1
          Gene Name:
          histidine triad nucleotide binding protein 1
          Gene Aliases:
          HINT, NMAN, PKCI-1, PRKCNH1
          Chromosome Location:
          Chr.5: 131159283 - 131165348 on Build GRCh38
          UniGene ID:
          Hs.483305
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531170

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HINT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005340.6 NP_005331.1 2 318
          NR_024610.2 2 318
          NR_024611.2 2 353
          NR_073488.1 2 353
          NR_134494.1 2 318
          NR_134495.1 2 318
          XM_017009410.1 XP_016864899.1 2 311
          GenBank mRNA AK026557.1 - 2 227
          BC001287.1 - 2 197 3458182
          BC007090.1 - 2 270 4250172
          BF967059.1 - 2 459
          BF970603.1 - 2 106
          BG393782.1 - 2 210
          BG527380.1 - 2 224
          BG708765.1 - 2 323
          CD639104.1 - 2 228
          CK002748.1 - 2 341
          CN290233.1 - 2 169
          CN290239.1 - 2 295
          CR457048.1 - 2 175
          DB125655.1 - 2 305
          DB289264.1 - 2 312
          U27143.1 - 2 218
          U51004.1 - 2 282
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA235754.1 - 2 261 Chr. 5 - 131162291 - 131165256 [-] on Build GRCh38
          GenBankmRNA BG504755.1 - 2 226 Chr. 5 - 131159285 - 131165220 [-] on Build GRCh38
          GenBankmRNA BG687022.1 - 2 224 Chr. 5 - 131159314 - 131165253 [-] on Build GRCh38
          GenBankmRNA BP231584.1 - 2 259 Chr. 5 - 131159329 - 131165254 [-] on Build GRCh38
          GenBankmRNA BG776401.1 - 2 224 Chr. 5 - 131159575 - 131165253 [-] on Build GRCh38
          GenBankmRNA AA203541.1 - 2 180 Chr. 5 - 131159404 - 131165211 [-] on Build GRCh38
          GenBankmRNA BG030650.1 - 2 270 Chr. 5 - 131159338 - 131165287 [-] on Build GRCh38
          GenBankmRNA BE874740.1 - 2 244 Chr. 5 - 131159292 - 131165239 [-] on Build GRCh38
          GenBankmRNA EL949508.1 - 2 205 Chr. 5 - 131159336 - 131165198 [-] on Build GRCh38
          GenBankmRNA BG776269.1 - 2 224 Chr. 5 - 131159467 - 131165253 [-] on Build GRCh38
          GenBankmRNA HY182087.1 - 2 237 Chr. 5 - 131159413 - 131165232 [-] on Build GRCh38
          GenBankmRNA BU533696.1 - 2 263 Chr. 5 - 131159285 - 131165255 [-] on Build GRCh38
          GenBankmRNA BF028892.1 - 2 193 Chr. 5 - 131159375 - 131165224 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotide phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          signal transduction
          purine ribonucleotide catabolic process
          positive regulation of calcium-mediated signaling
          intrinsic apoptotic signaling pathway by p53 class mediator
          nucleotide binding
          protein kinase C binding
          hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0