Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HINT1 SIRNA Assays ›
          Gene Symbol:
          HINT1
          Gene Name:
          histidine triad nucleotide binding protein 1
          Gene Aliases:
          HINT, NMAN, PKCI-1, PRKCNH1
          Chromosome Location:
          Chr.5: 131159283 - 131165348 on Build GRCh38
          UniGene ID:
          Hs.483305
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531171

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HINT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005340.6 NP_005331.1 1, 2 261
          NR_024610.2 1, 2 261
          NR_134494.1 1, 2 261
          NR_134495.1 1, 2 261
          XM_017009410.1 XP_016864899.1 1, 2 254
          GenBank mRNA AK026557.1 - 1, 2 170
          BC001287.1 - 1, 2 140 3458182
          BC007090.1 - 1, 2 213 4250172
          BF967059.1 - 1, 2 402
          BF970603.1 - 1, 2 49
          BG393782.1 - 1, 2 153
          BG527380.1 - 1, 2 167
          BG708765.1 - 1, 2 266
          BG942577.1 - 1, 2 253
          CN290233.1 - 1, 2 112
          CN290239.1 - 1, 2 238
          CR457048.1 - 1, 2 118
          DB125655.1 - 1, 2 248
          DB289264.1 - 1, 2 255
          U27143.1 - 1, 2 161
          U51004.1 - 1, 2 225
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG687022.1 - 1, 2 167 Chr. 5 - 131159314 - 131165253 [-] on Build GRCh38
          GenBankmRNA BG776401.1 - 1, 2 167 Chr. 5 - 131159575 - 131165253 [-] on Build GRCh38
          GenBankmRNA AA203541.1 - 1, 2 123 Chr. 5 - 131159404 - 131165211 [-] on Build GRCh38
          GenBankmRNA BG030650.1 - 1, 2 213 Chr. 5 - 131159338 - 131165287 [-] on Build GRCh38
          GenBankmRNA BG776269.1 - 1, 2 167 Chr. 5 - 131159467 - 131165253 [-] on Build GRCh38
          GenBankmRNA BF028892.1 - 1, 2 136 Chr. 5 - 131159375 - 131165224 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotide phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          signal transduction
          purine ribonucleotide catabolic process
          positive regulation of calcium-mediated signaling
          intrinsic apoptotic signaling pathway by p53 class mediator
          nucleotide binding
          protein kinase C binding
          hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0