Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103.3 | NP_001094.1 | 9 | 1022 | ||
NM_001278343.1 | NP_001265272.1 | 9 | 1022 | |||
NM_001278344.1 | NP_001265273.1 | 11 | 1219 | |||
GenBank mRNA | AB209521.1 | - | 11 | 1163 | ||
AK294906.1 | - | 7 | 533 | |||
AK297469.1 | - | 3 | 167 | |||
AK297618.1 | - | 2 | 104 | |||
AK297628.1 | - | 3 | 170 | |||
AK315250.1 | - | 9 | 1022 | |||
BC047901.2 | - | 9 | 954 | 6199155 | ||
BC051770.1 | - | 9 | 967 | 6198688 | ||
M86406.1 | - | 9 | 975 |