Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KDM1A SIRNA Assays ›
          Gene Symbol:
          KDM1A
          Gene Name:
          lysine demethylase 1A
          Gene Aliases:
          AOF2, BHC110, CPRF, KDM1, LSD1
          Chromosome Location:
          Chr.1: 23019443 - 23083691 on Build GRCh38
          UniGene ID:
          Hs.591518
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531634

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KDM1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001009999.2 NP_001009999.1 20 2537
          NM_015013.3 NP_055828.2 18 2465
          XM_005245786.2 XP_005245843.1 19 2530
          XM_006710472.3 XP_006710535.1 18 2488
          XM_006710473.2 XP_006710536.1 19 2548
          XM_006710474.3 XP_006710537.1 19 2548
          XM_017000716.1 XP_016856205.1 19 2530
          XM_017000717.1 XP_016856206.1 18 2470
          XM_017000718.1 XP_016856207.1 17 1954
          XM_017000719.1 XP_016856208.1 18 2080
          XM_017000720.1 XP_016856209.1 17 2050
          GenBank mRNA AB011173.1 - 18 2418
          AK128669.1 - 1 1437
          AK290328.1 - 18 1991
          AL833812.1 - 14 1587
          BC016639.1 - 16 1704
          BC025362.1 - 10 1130
          BC040194.2 - 20 2492 4815528
          BC048134.1 - 18 2386 5298150

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: EF688598.
          This siRNA also targets the following transcripts in Mouse: BC019417, BC059885, AK007763, AK129170.

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          protein demethylation
          blood coagulation
          cell proliferation
          regulation of double-strand break repair via homologous recombination
          regulation of primitive erythrocyte differentiation
          histone deacetylation
          pituitary gland development
          granulocyte differentiation
          negative regulation of protein binding
          histone H3-K9 demethylation
          positive regulation of histone ubiquitination
          cellular response to UV
          histone H3-K4 demethylation
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          positive regulation of erythrocyte differentiation
          positive regulation of megakaryocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hormone biosynthetic process
          positive regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of histone H3-K4 methylation
          negative regulation of histone H3-K9 methylation
          muscle cell development
          oxidation-reduction process
          cellular response to gamma radiation
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of cellular protein localization
          positive regulation of neural precursor cell proliferation
          positive regulation of stem cell proliferation
          RNA polymerase II transcription factor binding
          p53 binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          oxidoreductase activity
          enzyme binding
          ligand-dependent nuclear receptor transcription coactivator activity
          demethylase activity
          histone demethylase activity
          histone demethylase activity (H3-K4 specific)
          histone demethylase activity (H3-K9 specific)
          histone demethylase activity (H3-dimethyl-K4 specific)
          telomeric DNA binding
          MRF binding
          transcription regulatory region DNA binding
          flavin adenine dinucleotide binding
          androgen receptor binding
          telomeric repeat-containing RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0