Search
Search

EOGT| Gene Symbol: EOGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278689.1 | NP_001265618.1 | 18 | 4073 | ||
| NM_173654.2 | NP_775925.1 | 15 | 3821 | |||
| NR_103826.1 | 16 | 3862 | ||||
| XM_005264743.2 | XP_005264800.1 | 18 | 4287 | |||
| XM_011533599.1 | XP_011531901.1 | 19 | 4341 | |||
| XM_011533600.1 | XP_011531902.1 | 19 | 4194 | |||
| XM_011533602.1 | XP_011531904.1 | 19 | 4106 | |||
| XM_011533603.1 | XP_011531905.1 | 19 | 4338 | |||
| XM_017006204.1 | XP_016861693.1 | 19 | 4191 | |||
| XM_017006205.1 | XP_016861694.1 | 18 | 4052 | |||
| XM_017006206.1 | XP_016861695.1 | 18 | 3948 | |||
| XM_017006207.1 | XP_016861696.1 | 11 | 2787 | |||
| XM_017006209.1 | XP_016861698.1 | 9 | 2660 | |||
| GenBank mRNA | AK023140.1 | - | 1 | 1668 | ||
| AK091089.1 | - | 9 | 3051 | |||
| AK123213.1 | - | 6 | 2401 | |||
| AK126187.1 | - | 14 | 3146 | |||
| AL833467.1 | - | 4 | 2228 | |||
| BC028935.1 | - | 1 | 1363 | |||
| BC060887.1 | - | 15 | 3550 | 30347170 | ||
| BX640821.1 | - | 14 | 3194 | |||
| BX648818.1 | - | 18 | 3849 | |||