Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAD51 SIRNA Assays ›
          Gene Symbol:
          RAD51
          Gene Name:
          RAD51 recombinase
          Gene Aliases:
          BRCC5, FANCR, HRAD51, HsRad51, HsT16930, MRMV2, RAD51A, RECA
          Chromosome Location:
          Chr.15: 40694774 - 40732340 on Build GRCh38
          UniGene ID:
          Hs.631709
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531930

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAD51
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164269.1 NP_001157741.1 3 322
          NM_001164270.1 NP_001157742.1 3 477
          NM_002875.4 NP_002866.2 3 477
          NM_133487.3 NP_597994.3 3 477
          XM_006720626.3 XP_006720689.1 3 528
          XM_011521857.2 XP_011520159.1 3 227
          XM_011521858.2 XP_011520160.1 3 274
          XM_011521859.2 XP_011520161.1 3 411
          XM_011521860.2 XP_011520162.1 3 322
          XM_011521861.2 XP_011520163.1 3 227
          XM_011521862.2 XP_011520164.1 3 445
          GenBank mRNA AK131299.1 - 3 322
          AK291969.1 - 3 432
          AK303089.1 - 3 433
          AK313503.1 - 3 432
          AY425955.1 - 1 24
          BC001459.2 - 3 434 3139011
          BT019705.1 - 2 178
          CR536559.1 - 2 178
          D13804.1 - 3 389
          D14134.1 - 3 410
          EU362635.1 - 3 477

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          double-strand break repair via homologous recombination
          DNA recombinase assembly
          DNA synthesis involved in DNA repair
          strand displacement
          regulation of protein phosphorylation
          DNA unwinding involved in DNA replication
          DNA repair
          DNA recombination
          mitotic recombination
          cellular response to DNA damage stimulus
          meiotic nuclear division
          reciprocal meiotic recombination
          response to toxic substance
          response to X-ray
          regulation of double-strand break repair via homologous recombination
          telomere maintenance via telomere lengthening
          replication fork processing
          strand invasion
          response to drug
          positive regulation of DNA ligation
          protein homooligomerization
          chromosome organization involved in meiotic cell cycle
          cellular response to ionizing radiation
          cellular response to gamma radiation
          cellular response to hydroxyurea
          cellular response to cisplatin
          cellular response to camptothecin
          response to etoposide
          replication-born double-strand break repair via sister chromatid exchange
          mitotic recombination-dependent replication fork processing
          recombinase activity
          four-way junction DNA binding
          chromatin binding
          double-stranded DNA binding
          single-stranded DNA binding
          endodeoxyribonuclease activity
          protein binding
          ATP binding
          protein C-terminus binding
          identical protein binding
          single-stranded DNA-dependent ATPase activity
          DNA polymerase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0