Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207645.3 | NP_997528.2 | 2 | 593 | ||
XM_011542817.2 | XP_011541119.1 | 2 | 697 | |||
XM_011542818.2 | XP_011541120.1 | 2 | 575 | |||
GenBank mRNA | AK303693.1 | - | 2 | 593 | ||
BC035798.1 | - | 2 | 513 | 5735100 | ||
BC068577.1 | - | 2 | 566 | 4818568 |