Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SERPINB6 SIRNA Assays ›
Gene Symbol:
SERPINB6
Gene Name:
serpin family B member 6
Gene Aliases:
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3
Chromosome Location:
Chr.6: 2948159 - 2972165 on Build GRCh38
UniGene ID:
Hs.519523
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s532349
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SERPINB6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195291.2
NP_001182220.2
4
1080
NM_001271822.1
NP_001258751.1
3
505
NM_001271823.1
NP_001258752.1
3
603
NM_001271824.1
NP_001258753.1
3
987
NM_001271825.1
NP_001258754.1
3
589
NM_001297699.1
NP_001284628.1
3
1230
NM_001297700.1
NP_001284629.1
3
886
NM_004568.5
NP_004559.4
3
945
XM_011514672.1
XP_011512974.1
5
883
XM_011514673.1
XP_011512975.1
3
1037
XM_011514674.2
XP_011512976.1
3
622
XM_011514676.2
XP_011512978.1
3
432
XM_017010940.1
XP_016866429.1
4
444
XM_017010941.1
XP_016866430.1
2
2073
GenBank mRNA
AB209563.1
-
2
613
AF116727.1
-
1
349
AK027149.1
-
2
1771
AK057138.1
-
2
2304
AK094983.1
-
4
1080
AK098566.1
-
3
602
AK122895.1
-
1
1208
AK122961.1
-
2
1221
AK128637.1
-
1
2634
AK314578.1
-
3
664
BC001394.2
-
3
527
3051381
BC098564.1
-
3
383
30563105
BG499326.1
-
3
462
BI090002.1
-
3
587
BI259189.1
-
3
407
BM789272.1
-
3
568
BP325314.1
-
3
382
BP370484.1
-
3
438
BQ681973.1
-
3
392
BX375950.2
-
3
671
BX538343.1
-
3
4657
CX755337.1
-
3
508
CX782163.1
-
3
455
Z22658.1
-
3
376
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
sensory perception of sound
negative regulation of endopeptidase activity
cellular response to osmotic stress
protease binding
serine-type endopeptidase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0