Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAT1 SIRNA Assays ›
          Gene Symbol:
          STAT1
          Gene Name:
          signal transducer and activator of transcription 1
          Gene Aliases:
          CANDF7, IMD31A, IMD31B, IMD31C, ISGF-3, STAT91
          Chromosome Location:
          Chr.2: 190969036 - 191014250 on Build GRCh38
          UniGene ID:
          Hs.642990
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532488

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STAT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007315.3 NP_009330.1 13, 14 1514
          NM_139266.2 NP_644671.1 13, 14 1514
          XM_006712718.1 XP_006712781.1 12, 13 1307
          XM_017004783.1 XP_016860272.1 14, 15 1544
          GenBank mRNA AK096686.1 - 14, 15 1531
          AK292604.1 - 12, 13 1558
          AK315002.1 - 13, 14 1429
          BC002704.2 - 13, 14 1432 3627218
          BT007241.1 - 11, 12 1126
          CR749636.1 - 13, 14 1421
          GU211347.1 - 12, 13 1190
          GU211348.1 - 11, 12 1098
          M97935.1 - 12, 13 1322
          M97936.1 - 12, 13 1322

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of endothelial cell proliferation
          positive regulation of mesenchymal cell proliferation
          negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription, DNA-templated
          apoptotic process
          JAK-STAT cascade
          blood circulation
          macrophage derived foam cell differentiation
          negative regulation of angiogenesis
          tumor necrosis factor-mediated signaling pathway
          response to cytokine
          response to interferon-beta
          cellular response to interferon-beta
          regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to peptide hormone
          endothelial cell migration
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation by virus of viral protein levels in host cell
          positive regulation of smooth muscle cell proliferation
          response to cAMP
          defense response to virus
          interferon-gamma-mediated signaling pathway
          regulation of interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          regulation of type I interferon-mediated signaling pathway
          renal tubule development
          cellular response to organic cyclic compound
          metanephric mesenchymal cell proliferation involved in metanephros development
          metanephric mesenchymal cell differentiation
          negative regulation of metanephric nephron tubule epithelial cell differentiation
          cell-cell adhesion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          tumor necrosis factor receptor binding
          protein binding
          enzyme binding
          nuclear hormone receptor binding
          identical protein binding
          protein homodimerization activity
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0