Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000405.4 | NP_000396.2 | 3 | 508 | ||
NM_001167607.1 | NP_001161079.1 | 3 | 508 | |||
GenBank mRNA | AK225978.1 | - | 3 | 436 | ||
AK298867.1 | - | 3 | 375 | |||
AK312494.1 | - | 3 | 403 | |||
BC009273.2 | - | 3 | 372 | 4053681 | ||
BX473154.1 | - | 3 | 486 | |||
L01439.1 | - | 3 | 433 | |||
M76477.1 | - | 3 | 433 | |||
X16087.1 | - | 3 | 301 | |||
X61094.1 | - | 2 | 218 | |||
X61095.1 | - | 3 | 364 | |||
X62078.1 | - | 3 | 401 |