Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF2AK4 SIRNA Assays ›
          Gene Symbol:
          EIF2AK4
          Gene Name:
          eukaryotic translation initiation factor 2 alpha kinase 4
          Gene Aliases:
          GCN2, PVOD2
          Chromosome Location:
          Chr.15: 39934101 - 40035596 on Build GRCh38
          UniGene ID:
          Hs.656673
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532696

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EIF2AK4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013703.3 NP_001013725.2 34, 35 4635
          XM_005254392.2 XP_005254449.1 34, 35 4658
          XM_011521599.2 XP_011519901.1 34, 35 4658
          XM_011521600.2 XP_011519902.1 33, 34 4487
          XM_017022219.1 XP_016877708.1 33, 34 4487
          GenBank mRNA AB037759.1 - 30, 31 4101
          AJ243428.1 - 12, 13 1262
          AK027011.1 - 20, 21 2073
          AK074361.1 - 12, 13 1303
          AL137627.1 - 3, 4 189
          AL137676.1 - 10, 11 1082
          AL157497.1 - 23, 24 2432
          AL832907.1 - 28, 29 3895
          BC009350.2 - 20, 21 2105
          BX649048.1 - 17, 18 3480

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: FQ228781, CF108241.
          This siRNA also targets the following transcripts in Mouse: AF193343, AK129334, AF193342, BC072637, AF193344, and others.

          Panther Classification:

          Molecular Function -

          aminoacyl-tRNA synthetase non-receptor serine/threonine protein kinase translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          adaptive immune response
          T cell activation involved in immune response
          positive regulation of adaptive immune response
          regulation of translational initiation
          protein phosphorylation
          cell cycle arrest
          learning
          long-term memory
          regulation of translational initiation by eIF2 alpha phosphorylation
          viral translation
          negative regulation of translational initiation in response to stress
          negative regulation of CREB transcription factor activity
          cellular response to amino acid starvation
          cellular response to UV
          eiF2alpha phosphorylation in response to endoplasmic reticulum stress
          induction by virus of host autophagy
          negative regulation by host of viral genome replication
          negative regulation of neuron differentiation
          negative regulation of translational initiation
          protein autophosphorylation
          defense response to virus
          regulation of feeding behavior
          regulation of eIF2 alpha phosphorylation by amino acid starvation
          cellular response to cold
          positive regulation of translational initiation in response to starvation
          positive regulation of long-term synaptic potentiation
          neuron projection extension
          cellular response to leucine starvation
          tRNA binding
          protein serine/threonine kinase activity
          eukaryotic translation initiation factor 2alpha kinase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0