Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 14 | 2036 | ||
NM_001321185.1 | NP_001308114.1 | 14 | 1989 | |||
NM_007267.7 | NP_009198.4 | 14 | 1940 | |||
XM_005256995.1 | XP_005257052.1 | 14 | 2065 | |||
XM_011524255.1 | XP_011522557.1 | 13 | 1967 | |||
XM_011524256.1 | XP_011522558.1 | 13 | 1841 | |||
XM_011524257.2 | XP_011522559.1 | 8 | 1085 | |||
XM_017024107.1 | XP_016879596.1 | 13 | 1834 | |||
XM_017024108.1 | XP_016879597.1 | 13 | 1787 | |||
XM_017024109.1 | XP_016879598.1 | 12 | 1756 | |||
GenBank mRNA | AB002405.2 | - | 6 | 748 | ||
AK074065.1 | - | 14 | 1939 | |||
AK310734.1 | - | 13 | 2360 | |||
AY057379.1 | - | 13 | 1779 | |||
AY099356.1 | - | 15 | 1938 | |||
AY236496.1 | - | 13 | 1905 | |||
BC018346.1 | - | 7 | 904 | |||
BC023597.2 | - | 14 | 1920 | 4566166 | ||
BC035648.1 | - | 7 | 960 | 5575949 | ||
DQ104440.1 | - | 4 | 516 |