Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127198.2 | NP_001120670.1 | 20 | 2776 | ||
NM_001321185.1 | NP_001308114.1 | 20 | 2729 | |||
NM_007267.7 | NP_009198.4 | 20 | 2680 | |||
XM_005256995.1 | XP_005257052.1 | 20 | 2805 | |||
XM_011524255.1 | XP_011522557.1 | 19 | 2707 | |||
XM_011524256.1 | XP_011522558.1 | 19 | 2581 | |||
XM_011524257.2 | XP_011522559.1 | 14 | 1825 | |||
XM_011524258.1 | XP_011522560.1 | 11 | 1657 | |||
XM_017024107.1 | XP_016879596.1 | 19 | 2574 | |||
XM_017024108.1 | XP_016879597.1 | 19 | 2527 | |||
XM_017024109.1 | XP_016879598.1 | 18 | 2496 | |||
GenBank mRNA | AB002405.2 | - | 12 | 1488 | ||
AK074065.1 | - | 20 | 2679 | |||
AY099356.1 | - | 21 | 2678 | |||
AY099357.1 | - | 13 | 1625 | |||
AY236496.1 | - | 19 | 2645 | |||
BC018346.1 | - | 13 | 1644 | |||
BC023597.2 | - | 20 | 2660 | 4566166 | ||
BC035648.1 | - | 13 | 1700 | 5575949 |