Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000041.3 | NP_000032.1 | 3 | 186 | ||
NM_001302688.1 | NP_001289617.1 | 3 | 268 | |||
NM_001302689.1 | NP_001289618.1 | 3 | 117 | |||
NM_001302690.1 | NP_001289619.1 | 3 | 217 | |||
NM_001302691.1 | NP_001289620.1 | 3 | 201 | |||
GenBank mRNA | AK314898.1 | - | 3 | 139 | ||
BC003557.1 | - | 3 | 115 | 3355712 | ||
BI550631.1 | - | 3 | 163 | |||
BM042032.1 | - | 3 | 201 | |||
BU848796.1 | - | 153 | ||||
CA771535.1 | - | 2 | 335 | |||
DB486513.1 | - | 3 | 118 | |||
HY056112.1 | - | 3 | 186 | |||
HY074384.1 | - | 3 | 117 | |||
K00396.1 | - | 3 | 130 | |||
KU177910.1 | - | 2 | 70 | |||
KU177911.1 | - | 2 | 70 | |||
KU177912.1 | - | 2 | 70 | |||
KU177913.1 | - | 2 | 70 | |||
M12529.1 | - | 3 | 131 | |||
Z70760.1 | - | 2 | 107 |