Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUNX2 SIRNA Assays ›
          Gene Symbol:
          RUNX2
          Gene Name:
          runt related transcription factor 2
          Gene Aliases:
          AML3, CBF-alpha-1, CBFA1, CCD, CCD1, CLCD, OSF-2, OSF2, PEA2aA, PEBP2aA
          Chromosome Location:
          Chr.6: 45328142 - 45664032 on Build GRCh38
          UniGene ID:
          Hs.535845
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532996

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUNX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001015051.3 NP_001015051.3 5 863
          NM_001024630.3 NP_001019801.3 5 863
          NM_001278478.1 NP_001265407.1 3 611
          XM_006715232.1 XP_006715295.1 3 614
          XM_011514960.2 XP_011513262.1 5 1038
          XM_011514961.2 XP_011513263.1 5 1038
          XM_011514962.2 XP_011513264.1 5 1038
          XM_011514963.2 XP_011513265.1 5 1038
          XM_011514964.2 XP_011513266.1 5 1038
          XM_011514965.2 XP_011513267.1 5 1038
          XM_011514966.2 XP_011513268.1 3 1681
          XM_017011391.1 XP_016866880.1 5 1073
          XM_017011392.1 XP_016866881.1 3 614
          XM_017011393.1 XP_016866882.1 5 1038
          XM_017011394.1 XP_016866883.1 5 1038
          XM_017011395.1 XP_016866884.1 5 1038
          XM_017011396.1 XP_016866885.1 3 614
          GenBank mRNA BC108919.1 - 3 593
          BC108920.1 - 3 597
          L40992.1 - 3 476

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Runt transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          osteoblast differentiation
          endochondral ossification
          osteoblast fate commitment
          chondrocyte differentiation
          chondrocyte development
          osteoblast development
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          hemopoiesis
          neuron differentiation
          T cell differentiation
          BMP signaling pathway
          positive regulation of chondrocyte differentiation
          embryonic forelimb morphogenesis
          regulation of fibroblast growth factor receptor signaling pathway
          odontogenesis of dentin-containing tooth
          regulation of odontogenesis of dentin-containing tooth
          cellular response to fibroblast growth factor stimulus
          regulation of cell differentiation
          positive regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          cell maturation
          embryonic cranial skeleton morphogenesis
          stem cell differentiation
          cellular response to BMP stimulus
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          ATP binding
          protein domain specific binding
          histone deacetylase binding
          bHLH transcription factor binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0