Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEF2C SIRNA Assays ›
Gene Symbol:
MEF2C
Gene Name:
myocyte enhancer factor 2C
Gene Aliases:
C5DELq14.3, DEL5q14.3
Chromosome Location:
Chr.5: 88718241 - 88904105 on Build GRCh38
UniGene ID:
Hs.649965
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533102
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEF2C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001131005.2
NP_001124477.1
8
1159
NM_001193347.1
NP_001180276.1
9
1219
NM_001193348.1
NP_001180277.1
5
731
NM_001193349.1
NP_001180278.1
5
731
NM_001193350.1
NP_001180279.1
7
1387
NM_001308002.1
NP_001294931.1
6
875
NM_002397.4
NP_002388.2
7
1390
XM_005248511.2
XP_005248568.1
7
924
XM_006714619.2
XP_006714682.1
8
1246
XM_006714625.3
XP_006714688.1
8
1277
XM_011543396.2
XP_011541698.1
7
975
XM_011543397.2
XP_011541699.1
7
1156
XM_011543400.1
XP_011541702.1
7
1162
XM_011543401.1
XP_011541703.1
7
1162
XM_017009475.1
XP_016864964.1
7
1217
XM_017009476.1
XP_016864965.1
7
1223
XM_017009477.1
XP_016864966.1
7
1162
XM_017009478.1
XP_016864967.1
7
1217
XM_017009479.1
XP_016864968.1
7
1223
XM_017009480.1
XP_016864969.1
8
1246
XM_017009481.1
XP_016864970.1
7
924
XM_017009482.1
XP_016864971.1
7
1223
XM_017009483.1
XP_016864972.1
7
1217
GenBank mRNA
AK307883.1
-
6
732
AK312472.1
-
7
1095
AL833268.1
-
9
1167
AL833274.1
-
8
1108
BC026341.1
-
7
1162
FM163484.2
-
5
592
FM180475.1
-
5
592
L08895.1
-
7
1137
S57212.1
-
1396
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
blood vessel development
osteoblast differentiation
neuron migration
B cell homeostasis
heart looping
endochondral ossification
blood vessel remodeling
chondrocyte differentiation
germinal center formation
regulation of germinal center formation
primary heart field specification
secondary heart field specification
outflow tract morphogenesis
sinoatrial valve morphogenesis
cardiac ventricle formation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
humoral immune response
nervous system development
heart development
muscle organ development
skeletal muscle tissue development
muscle cell fate determination
learning or memory
response to virus
positive regulation of gene expression
negative regulation of gene expression
positive regulation of alkaline phosphatase activity
neural crest cell differentiation
cardiac muscle hypertrophy in response to stress
myotube differentiation
neuron differentiation
platelet formation
monocyte differentiation
negative regulation of ossification
melanocyte differentiation
positive regulation of bone mineralization
positive regulation of B cell proliferation
cellular response to drug
skeletal muscle cell differentiation
cellular response to trichostatin A
B cell proliferation
regulation of neuron apoptotic process
negative regulation of neuron apoptotic process
regulation of megakaryocyte differentiation
positive regulation of myoblast differentiation
positive regulation of neuron differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of neurotransmitter secretion
regulation of synaptic plasticity
positive regulation of skeletal muscle tissue development
neuron development
cell morphogenesis involved in neuron differentiation
embryonic viscerocranium morphogenesis
negative regulation of epithelial cell proliferation
B cell receptor signaling pathway
smooth muscle cell differentiation
positive regulation of muscle cell differentiation
regulation of synapse assembly
regulation of synaptic transmission, glutamatergic
ventricular cardiac muscle cell differentiation
palate development
regulation of synaptic activity
positive regulation of cardiac muscle cell proliferation
excitatory postsynaptic potential
regulation of sarcomere organization
cartilage morphogenesis
regulation of dendritic spine development
renal tubule morphogenesis
cellular response to lipopolysaccharide
cellular response to calcium ion
cellular response to parathyroid hormone stimulus
cellular response to fluid shear stress
cellular response to transforming growth factor beta stimulus
positive regulation of cell proliferation in bone marrow
glomerulus morphogenesis
nephron tubule epithelial cell differentiation
positive regulation of protein homodimerization activity
positive regulation of macrophage apoptotic process
regulation of N-methyl-D-aspartate selective glutamate receptor activity
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
positive regulation of cardiac muscle cell differentiation
positive regulation of behavioral fear response
epithelial cell proliferation involved in renal tubule morphogenesis
positive regulation of skeletal muscle cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
AT DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
activating transcription factor binding
miRNA binding
histone deacetylase binding
transcription regulatory region DNA binding
protein heterodimerization activity
HMG box domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0