Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DYSF SIRNA Assays ›
Gene Symbol:
DYSF
Gene Name:
dysferlin
Gene Aliases:
FER1L1, LGMD2B, MMD1
Chromosome Location:
Chr.2: 71453155 - 71686763 on Build GRCh38
UniGene ID:
Hs.252180
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533194
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DYSF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130455.1
NP_001123927.1
46
5310
NM_001130976.1
NP_001124448.1
45
5500
NM_001130977.1
NP_001124449.1
46
5563
NM_001130978.1
NP_001124450.1
47
5605
NM_001130979.1
NP_001124451.1
47
5635
NM_001130980.1
NP_001124452.1
46
5593
NM_001130981.1
NP_001124453.1
47
5656
NM_001130982.1
NP_001124454.1
47
5403
NM_001130983.1
NP_001124455.1
47
5373
NM_001130984.1
NP_001124456.1
46
5331
NM_001130985.1
NP_001124457.1
46
5361
NM_001130986.1
NP_001124458.1
45
5268
NM_001130987.1
NP_001124459.1
47
5424
NM_003494.3
NP_003485.1
46
5542
XM_005264584.4
XP_005264641.1
48
5481
XM_005264585.4
XP_005264642.1
48
6166
GenBank mRNA
AF075575.1
-
46
5539
AK074104.1
-
32
3812
AK074649.1
-
6
679
AK295004.1
-
17
1999
AK303364.1
-
13
1613
BC171803.1
-
50
5657
DQ267935.1
-
46
5310
EU515155.1
-
47
5536
EU515156.1
-
45
5401
EU515157.1
-
47
5506
EU515158.1
-
46
5494
EU515159.1
-
46
5464
EU515160.1
-
47
5557
EU515161.1
-
47
5403
EU515162.1
-
45
5268
EU515163.1
-
47
5373
EU515164.1
-
46
5361
EU515165.1
-
46
5331
EU515166.1
-
47
5424
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
plasma membrane repair
vesicle fusion
muscle contraction
calcium ion binding
protein binding
phospholipid binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0