Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GATA3 SIRNA Assays ›
          Gene Symbol:
          GATA3
          Gene Name:
          GATA binding protein 3
          Gene Aliases:
          HDR, HDRS
          Chromosome Location:
          Chr.10: 8045421 - 8075201 on Build GRCh38
          UniGene ID:
          Hs.524134
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533215

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GATA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001002295.1 NP_001002295.1 6 1642
          NM_002051.2 NP_002042.1 6 1639
          XM_005252442.2 XP_005252499.1 6 1624
          XM_005252443.4 XP_005252500.1 6 1549
          GenBank mRNA AK298213.1 - 5 869
          BC003070.2 - 6 1533 3504200
          BC006793.1 - 6 1644 2985843
          M69106.1 - 5 1204
          X55037.1 - 5 1206
          X55122.1 - 5 1283
          X58072.1 - 5 1233

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          cell fate determination
          neuron migration
          type IV hypersensitivity
          kidney development
          mesonephros development
          lens development in camera-type eye
          pro-T cell differentiation
          aortic valve morphogenesis
          cardiac right ventricle morphogenesis
          ventricular septum development
          chromatin remodeling
          transcription from RNA polymerase II promoter
          defense response
          humoral immune response
          signal transduction
          axon guidance
          heart development
          blood coagulation
          negative regulation of cell proliferation
          male gonad development
          response to virus
          anatomical structure morphogenesis
          post-embryonic development
          organ morphogenesis
          positive regulation of signal transduction
          response to gamma radiation
          positive regulation of endothelial cell migration
          regulation of neuron projection development
          phosphatidylinositol 3-kinase signaling
          erythrocyte differentiation
          TOR signaling
          negative regulation of interferon-gamma production
          negative regulation of interleukin-2 production
          positive regulation of interleukin-4 production
          negative regulation of mammary gland epithelial cell proliferation
          embryonic hemopoiesis
          cellular response to interferon-alpha
          ureter maturation
          parathyroid hormone secretion
          regulation of cytokine biosynthetic process
          norepinephrine biosynthetic process
          inner ear morphogenesis
          response to drug
          regulation of CD4-positive, alpha-beta T cell differentiation
          regulation of neuron apoptotic process
          ear development
          response to estrogen
          thymic T cell selection
          T-helper 2 cell differentiation
          innate immune response
          cell fate commitment
          response to ethanol
          positive regulation of T cell differentiation
          negative regulation of fat cell differentiation
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          sympathetic nervous system development
          thymus development
          digestive tract development
          developmental growth
          anatomical structure formation involved in morphogenesis
          negative regulation of inflammatory response
          T cell receptor signaling pathway
          regulation of histone H3-K4 methylation
          positive regulation of protein kinase B signaling
          parathyroid gland development
          pharyngeal system development
          uterus development
          mesenchymal to epithelial transition
          mast cell differentiation
          ureteric bud formation
          regulation of histone H3-K27 methylation
          canonical Wnt signaling pathway involved in metanephric kidney development
          cellular response to interleukin-4
          cellular response to tumor necrosis factor
          positive regulation of histone H3-K14 acetylation
          otic vesicle development
          cellular response to BMP stimulus
          positive regulation of ureteric bud formation
          nephric duct morphogenesis
          nephric duct formation
          regulation of nephron tubule epithelial cell differentiation
          interleukin-4 secretion
          interferon-gamma secretion
          lymphocyte migration
          negative regulation of DNA demethylation
          regulation of establishment of cell polarity
          negative regulation of cell motility
          negative regulation of endothelial cell apoptotic process
          positive regulation of T-helper 2 cell cytokine production
          negative regulation of cell proliferation involved in mesonephros development
          positive regulation of thyroid hormone generation
          positive regulation of histone H3-K9 acetylation
          positive regulation of interleukin-5 secretion
          positive regulation of interleukin-13 secretion
          positive regulation of transcription regulatory region DNA binding
          regulation of cellular response to X-ray
          negative regulation of fibroblast growth factor receptor signaling pathway involved in ureteric bud formation
          negative regulation of glial cell-derived neurotrophic factor receptor signaling pathway involved in ureteric bud formation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          nucleic acid binding transcription factor activity
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          interleukin-2 receptor binding
          protein binding
          transcription factor binding
          zinc ion binding
          transcription regulatory region DNA binding
          protein dimerization activity
          E-box binding
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0