Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHRM3 SIRNA Assays ›
          Gene Symbol:
          CHRM3
          Gene Name:
          cholinergic receptor muscarinic 3
          Gene Aliases:
          EGBRS, HM3, PBS
          Chromosome Location:
          Chr.1: 239386565 - 239915450 on Build GRCh38
          UniGene ID:
          Hs.155736
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533393

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHRM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000740.2 NP_000731.1 5 1041
          XM_005273032.3 XP_005273089.1 6 1346
          XM_011544041.2 XP_011542343.1 7 1445
          XM_011544043.2 XP_011542345.1 9 1778
          XM_011544044.2 XP_011542346.1 8 1549
          XM_011544045.2 XP_011542347.1 8 1674
          XM_011544046.2 XP_011542348.1 6 1382
          XM_011544047.2 XP_011542349.1 6 1318
          XM_017000152.1 XP_016855641.1 8 4371
          XM_017000153.1 XP_016855642.1 7 1449
          XM_017000154.1 XP_016855643.1 6 1143
          XM_017000155.1 XP_016855644.1 7 1486
          XM_017000156.1 XP_016855645.1 7 1450
          XM_017000157.1 XP_016855646.1 5 1051
          XM_017000158.1 XP_016855647.1 5 1283
          XM_017000159.1 XP_016855648.1 5 1255
          XM_017000160.1 XP_016855649.1 4 2105
          XM_017000161.1 XP_016855650.1 6 1387
          XM_017000162.1 XP_016855651.1 5 1219
          XM_017000163.1 XP_016855652.1 4 5081
          GenBank mRNA AF279779.1 - 5 1040
          AF385589.1 - 1 262
          AF498917.1 - 1 262
          AK074575.1 - 1 419
          BC096844.1 - 1 263
          BC121026.2 - 2 405 40117429

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of vascular smooth muscle contraction
          cellular protein modification process
          smooth muscle contraction
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled acetylcholine receptor signaling pathway
          phospholipase C-activating G-protein coupled acetylcholine receptor signaling pathway
          G-protein coupled acetylcholine receptor signaling pathway
          synaptic transmission, cholinergic
          nervous system development
          cell proliferation
          positive regulation of smooth muscle contraction
          saliva secretion
          phosphatidylinositol phospholipase C activity
          receptor activity
          protein binding
          drug binding
          G-protein coupled acetylcholine receptor activity
          acetylcholine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0