Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRIM2 SIRNA Assays ›
          Gene Symbol:
          TRIM2
          Gene Name:
          tripartite motif containing 2
          Gene Aliases:
          CMT2R, RNF86
          Chromosome Location:
          Chr.4: 153152281 - 153339322 on Build GRCh38
          UniGene ID:
          Hs.435711
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533397

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRIM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130067.1 NP_001123539.1 11 2230
          NM_015271.4 NP_056086.2 11 2227
          XM_006714157.1 XP_006714220.1 11 2239
          XM_006714158.1 XP_006714221.1 11 2204
          XM_006714159.3 XP_006714222.1 11 3050
          XM_006714160.1 XP_006714223.1 12 3004
          XM_006714161.1 XP_006714224.1 12 3001
          XM_006714162.1 XP_006714225.1 12 2721
          XM_006714165.3 XP_006714228.1 9 2432
          XM_011531794.1 XP_011530096.1 11 2318
          XM_011531795.1 XP_011530097.1 12 2724
          XM_011531796.2 XP_011530098.1 11 2254
          XM_011531798.2 XP_011530100.1 11 2359
          XM_017007943.1 XP_016863432.1 11 2192
          XM_017007944.1 XP_016863433.1 12 2992
          XM_017007945.1 XP_016863434.1 12 2989
          XM_017007946.1 XP_016863435.1 11 3035
          XM_017007947.1 XP_016863436.1 11 2306
          XM_017007948.1 XP_016863437.1 11 2260
          XM_017007949.1 XP_016863438.1 12 2709
          XM_017007950.1 XP_016863439.1 12 2712
          XM_017007951.1 XP_016863440.1 11 2406
          XM_017007952.1 XP_016863441.1 11 2414
          XM_017007953.1 XP_016863442.1 9 1770
          XM_017007954.1 XP_016863443.1 12 2284
          XM_017007955.1 XP_016863444.1 12 2277
          GenBank mRNA AB011089.1 - 11 2175
          AF220018.1 - 11 2175
          AL110234.1 - 4 487
          BC005016.1 - 6 747
          BC011052.1 - 11 2230 4156234

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          regulation of neuron apoptotic process
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0