Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOD2 SIRNA Assays ›
          Gene Symbol:
          NOD2
          Gene Name:
          nucleotide binding oligomerization domain containing 2
          Gene Aliases:
          ACUG, BLAU, CARD15, CD, CLR16.3, IBD1, NLRC2, NOD2B, PSORAS1
          Chromosome Location:
          Chr.16: 50693581 - 50733077 on Build GRCh38
          UniGene ID:
          Hs.592072
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533470

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NOD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001293557.1 NP_001280486.1 3 1389
          NM_022162.2 NP_071445.1 4 1429
          XM_005256084.3 XP_005256141.1 4 1333
          XM_006721242.3 XP_006721305.1 4 1333
          XM_006721243.3 XP_006721306.1 4 1333
          XM_011523259.2 XP_011521561.1 2 1145
          XM_011523260.2 XP_011521562.1 4 1333
          XM_011523261.1 XP_011521563.1 4 1333
          XM_017023535.1 XP_016879024.1 2 2703
          XM_017023536.1 XP_016879025.1 3 860
          XM_017023537.1 XP_016879026.1 2 754
          XM_017023538.1 XP_016879027.1 3 855
          GenBank mRNA AF178930.1 - 4 1429

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          cytokine production involved in immune response
          cytokine secretion involved in immune response
          positive regulation of dendritic cell antigen processing and presentation
          positive regulation of type 2 immune response
          defense response
          JNK cascade
          detection of biotic stimulus
          detection of bacterium
          maintenance of gastrointestinal epithelium
          response to muramyl dipeptide
          detection of muramyl dipeptide
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-17 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of stress-activated MAPK cascade
          activation of MAPK activity involved in innate immune response
          intracellular signal transduction
          defense response to bacterium
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of phosphatidylinositol 3-kinase activity
          innate immune response
          positive regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of gamma-delta T cell activation
          positive regulation of epithelial cell proliferation
          positive regulation of interleukin-1 beta secretion
          regulation of inflammatory response
          positive regulation of B cell activation
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          positive regulation of oxidoreductase activity
          positive regulation of nitric-oxide synthase biosynthetic process
          positive regulation of prostaglandin-endoperoxide synthase activity
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          cellular response to muramyl dipeptide
          positive regulation of cytokine production involved in inflammatory response
          positive regulation of NIK/NF-kappaB signaling
          negative regulation of macrophage apoptotic process
          positive regulation of prostaglandin-E synthase activity
          actin binding
          protein binding
          ATP binding
          enzyme binding
          protein kinase binding
          Hsp70 protein binding
          muramyl dipeptide binding
          peptidoglycan binding
          CARD domain binding
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0