Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCTN1 SIRNA Assays ›
Gene Symbol:
TCTN1
Gene Name:
tectonic family member 1
Gene Aliases:
JBTS13, TECT1
Chromosome Location:
Chr.12: 110614027 - 110649130 on Build GRCh38
UniGene ID:
Hs.211511
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533594
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001082537.2
NP_001076006.1
8
1123
NM_001082538.2
NP_001076007.1
8
1123
NM_001173975.2
NP_001167446.1
8
1184
NM_001173976.1
NP_001167447.1
9
1199
NM_001319680.1
NP_001306609.1
8
1123
NM_001319681.1
NP_001306610.1
9
1296
NM_024549.5
NP_078825.2
8
1081
NR_135088.1
9
1479
XM_005253934.3
XP_005253991.1
8
1027
XM_005253935.3
XP_005253992.1
8
1027
XM_006719594.2
XP_006719657.1
8
1088
XM_006719595.2
XP_006719658.1
9
1194
XM_006719596.2
XP_006719659.1
9
1217
XM_006719597.3
XP_006719660.1
7
914
XM_006719598.2
XP_006719661.1
9
1187
XM_006719599.2
XP_006719662.1
8
992
XM_006719600.2
XP_006719663.1
7
931
XM_011538733.2
XP_011537035.1
8
985
XM_011538734.2
XP_011537036.1
7
982
XM_011538735.1
XP_011537037.1
8
1027
XM_011538737.2
XP_011537039.1
8
1027
XM_011538738.2
XP_011537040.1
8
1027
XM_017019964.1
XP_016875453.1
8
1088
XM_017019965.1
XP_016875454.1
9
1306
XM_017019966.1
XP_016875455.1
9
1353
XM_017019967.1
XP_016875456.1
9
1061
XM_017019968.1
XP_016875457.1
8
1150
XM_017019969.1
XP_016875458.1
8
950
GenBank mRNA
AK024780.1
-
9
1097
AK055891.1
-
8
995
AK092775.1
-
8
1085
AK301732.1
-
9
1161
AY358184.1
-
8
986
BC040113.1
-
8
971
6054789
BC044885.1
-
9
1341
BC062611.1
-
8
974
5184202
DQ278868.1
-
8
1048
Back To Top
More Information
Gene Ontology Categories:
Function(s)
in utero embryonic development
neural tube formation
regulation of smoothened signaling pathway
somatic motor neuron differentiation
telencephalon development
dorsal/ventral neural tube patterning
central nervous system interneuron axonogenesis
cilium morphogenesis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0