Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GCK SIRNA Assays ›
          Gene Symbol:
          GCK
          Gene Name:
          glucokinase
          Gene Aliases:
          FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
          Chromosome Location:
          Chr.7: 44143213 - 44189439 on Build GRCh38
          UniGene ID:
          Hs.1270
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533608

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GCK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000162.3 NP_000153.1 7, 8 1339
          NM_033507.1 NP_277042.1 7, 8 1040
          NM_033508.1 NP_277043.1 8, 9 1164
          XM_017011966.1 XP_016867455.1 7, 8 1355
          GenBank mRNA AK122876.1 - 7, 8 1034
          BC001890.1 - 7, 8 1339 3536582
          BT007062.1 - 7, 8 869
          EU008605.1 - 7, 8 869
          M69051.1 - 8, 9 1151
          M88011.1 - 7, 8 1198
          M90299.1 - 7, 8 1327

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular glucose homeostasis
          glycogen biosynthetic process
          fructose 2,6-bisphosphate metabolic process
          glycolytic process
          regulation of glycolytic process
          NADP metabolic process
          positive regulation of cytosolic calcium ion concentration
          regulation of glucose transport
          glucose transport
          second-messenger-mediated signaling
          positive regulation of insulin secretion
          negative regulation of epinephrine secretion
          cellular response to insulin stimulus
          cellular response to glucose starvation
          glucose homeostasis
          regulation of potassium ion transport
          cellular response to leptin stimulus
          negative regulation of gluconeogenesis
          positive regulation of glycogen biosynthetic process
          positive regulation of glycolytic process
          carbohydrate phosphorylation
          regulation of insulin secretion
          glucose 6-phosphate metabolic process
          detection of glucose
          lipid homeostasis
          canonical glycolysis
          calcium ion import
          magnesium ion binding
          glucokinase activity
          protein binding
          ATP binding
          glucose binding
          fructokinase activity
          mannokinase activity
          protein phosphatase binding
          ADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0