Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS4 SIRNA Assays ›
Gene Symbol:
HPS4
Gene Name:
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases:
BLOC3S2, LE
Chromosome Location:
Chr.22: 26443613 - 26483863 on Build GRCh38
UniGene ID:
Hs.474436
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533762
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022081.5
NP_071364.4
14
3009
NM_152841.2
NP_690054.1
12
2626
NR_073135.1
14
3027
NR_073136.1
13
2789
XM_006724353.3
XP_006724416.1
15
3054
XM_006724354.3
XP_006724417.1
15
2959
XM_006724360.3
XP_006724423.1
10
2237
XM_011530485.1
XP_011528787.1
16
3132
XM_011530486.2
XP_011528788.1
16
3037
XM_011530487.2
XP_011528789.1
17
3242
XM_011530488.2
XP_011528790.1
16
3040
XM_011530489.2
XP_011528791.1
17
3143
XM_011530490.2
XP_011528792.1
15
3078
XM_011530493.2
XP_011528795.1
14
2890
XM_011530494.2
XP_011528796.1
14
2428
XM_011530495.2
XP_011528797.1
8
1934
XM_011530496.2
XP_011528798.1
9
1769
XM_017029042.1
XP_016884531.1
14
2905
XM_017029043.1
XP_016884532.1
15
3110
XM_017029044.1
XP_016884533.1
14
2908
XM_017029046.1
XP_016884535.1
14
3568
XM_017029047.1
XP_016884536.1
13
2836
XM_017029050.1
XP_016884539.1
12
2758
XM_017029051.1
XP_016884540.1
12
2663
XM_017029052.1
XP_016884541.1
15
3163
XM_017029053.1
XP_016884542.1
7
2522
XM_017029054.1
XP_016884543.1
14
3109
XM_017029055.1
XP_016884544.1
14
2963
XM_017029056.1
XP_016884545.1
15
3176
XM_017029057.1
XP_016884546.1
15
3025
XM_017029058.1
XP_016884547.1
15
3059
XM_017029059.1
XP_016884548.1
15
2934
XM_017029060.1
XP_016884549.1
15
2933
XM_017029061.1
XP_016884550.1
15
3134
XM_017029062.1
XP_016884551.1
13
2909
XM_017029063.1
XP_016884552.1
15
3043
XM_017029064.1
XP_016884553.1
13
2792
GenBank mRNA
AB051454.1
-
13
5199
AK095406.1
-
4
1546
AK097529.1
-
2
414
AK123691.1
-
9
2736
AL365512.1
-
12
2530
AL713795.1
-
14
2958
AY043416.1
-
13
2529
BC028625.1
-
14
3023
BC050285.1
-
14
3000
BC065030.1
-
13
2731
6148557
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
lysosome organization
blood coagulation
hemostasis
melanocyte differentiation
positive regulation of eye pigmentation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein binding
protein homodimerization activity
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0