Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STAT5B SIRNA Assays ›
          Gene Symbol:
          STAT5B
          Gene Name:
          signal transducer and activator of transcription 5B
          Gene Aliases:
          STAT5
          Chromosome Location:
          Chr.17: 42199177 - 42276460 on Build GRCh38
          UniGene ID:
          Hs.595276
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533811

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STAT5B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012448.3 NP_036580.2 2 268
          XM_005257626.4 XP_005257683.1 2 322
          XM_017024977.1 XP_016880466.1 2 244
          GenBank mRNA AB208920.1 - 2 247
          AK308163.1 - 2 156
          BC020868.1 - 2 268
          BC065227.1 - 2 268 5553426
          U47686.1 - 2 245
          U48730.2 - 1 110

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          allantoin metabolic process
          luteinization
          response to hypoxia
          natural killer cell differentiation
          liver development
          citrate metabolic process
          2-oxoglutarate metabolic process
          succinate metabolic process
          oxaloacetate metabolic process
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          isoleucine metabolic process
          valine metabolic process
          creatine metabolic process
          fatty acid metabolic process
          acute-phase response
          JAK-STAT cascade
          female pregnancy
          lactation
          peptidyl-tyrosine phosphorylation
          regulation of steroid metabolic process
          taurine metabolic process
          lipid storage
          regulation of epithelial cell differentiation
          response to estradiol
          response to lipopolysaccharide
          positive regulation of natural killer cell proliferation
          positive regulation of natural killer cell differentiation
          cellular response to hormone stimulus
          T cell differentiation in thymus
          prolactin signaling pathway
          regulation of multicellular organism growth
          positive regulation of multicellular organism growth
          positive regulation of activated T cell proliferation
          progesterone metabolic process
          T cell homeostasis
          negative regulation of apoptotic process
          positive regulation of interleukin-2 biosynthetic process
          response to ethanol
          positive regulation of B cell differentiation
          positive regulation of gamma-delta T cell differentiation
          negative regulation of erythrocyte differentiation
          positive regulation of erythrocyte differentiation
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of natural killer cell mediated cytotoxicity
          creatinine metabolic process
          development of secondary female sexual characteristics
          development of secondary male sexual characteristics
          Peyer's patch development
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of cellular component movement
          JAK-STAT cascade involved in growth hormone signaling pathway
          response to interleukin-2
          response to interleukin-4
          response to interleukin-15
          cellular response to growth factor stimulus
          cellular response to epidermal growth factor stimulus
          mast cell migration
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein tyrosine kinase activity
          signal transducer activity
          protein binding
          protein phosphatase binding
          glucocorticoid receptor binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cqs7j:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0