Search
Search

FUT2| Gene Symbol: FUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000511.5 | NP_000502.4 | 2 | 1292 | ||
| NM_001097638.2 | NP_001091107.1 | 2 | 1282 | |||
| GenBank mRNA | BC001899.1 | - | 2 | 1232 | ||
| BC121066.1 | - | 2 | 1214 | |||
| D87942.1 | - | 2 | 1282 | |||