Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136024.3 | NP_001129496.1 | 7 | 2492 | ||
NM_001291867.1 | NP_001278796.1 | 7 | 3087 | |||
NM_001291868.1 | NP_001278797.1 | 6 | 2429 | |||
NM_198270.3 | NP_938011.1 | 6 | 3024 | |||
XM_011545528.2 | XP_011543830.1 | 4 | 2636 | |||
GenBank mRNA | AK302437.1 | - | 7 | 3273 | ||
AY436752.1 | - | 6 | 3024 | |||
AY456992.1 | - | 7 | 2492 | |||
AY456993.1 | - | 6 | 3147 | |||
BC136415.1 | - | 7 | 2813 | 9020403 | ||
BC171763.1 | - | 7 | 2813 | 9054416 | ||
CR936788.1 | - | 6 | 2198 | |||
GQ988776.1 | - | 7 | 2752 |