Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNAAF1 SIRNA Assays ›
          Gene Symbol:
          DNAAF1
          Gene Name:
          dynein axonemal assembly factor 1
          Gene Aliases:
          CILD13, LRRC50, ODA7
          Chromosome Location:
          Chr.16: 84145260 - 84178761 on Build GRCh38
          UniGene ID:
          Hs.310164
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533845

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DNAAF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318756.1 NP_001305685.1 5 1094
          NM_178452.5 NP_848547.4 9 1796
          XM_006721129.2 XP_006721192.1 9 1796
          XM_011522853.2 XP_011521155.1 9 1844
          XM_011522854.2 XP_011521156.1 9 1844
          XM_011522855.2 XP_011521157.1 9 1844
          XM_011522857.2 XP_011521159.1 9 1844
          XM_011522858.2 XP_011521160.1 9 1844
          XM_017022918.1 XP_016878407.1 9 1796
          XM_017022919.1 XP_016878408.1 9 2152
          XM_017022920.1 XP_016878409.1 5 1292
          XM_017022921.1 XP_016878410.1 5 1244
          XM_017022922.1 XP_016878411.1 5 1244
          GenBank mRNA AK057238.1 - 10 1560
          AK058059.1 - 5 1053
          AK295990.1 - 5 1094
          AL833327.1 - 9 1767
          AL833328.1 - 9 1767
          AL833336.1 - 9 1767
          BC024009.2 - 9 1739 4822973
          BX648657.1 - 9 1769

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein-binding activity modulator phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          heart looping
          cilium movement
          regulation of cilium beat frequency
          lung development
          determination of pancreatic left/right asymmetry
          outer dynein arm assembly
          inner dynein arm assembly
          motile cilium assembly
          cilium morphogenesis
          epithelial cilium movement involved in determination of left/right asymmetry
          left/right pattern formation
          axonemal dynein complex assembly
          determination of digestive tract left/right asymmetry
          determination of liver left/right asymmetry
          dynein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0