Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD46 SIRNA Assays ›
          Gene Symbol:
          CD46
          Gene Name:
          CD46 molecule
          Gene Aliases:
          AHUS2, MCP, MIC10, TLX, TRA2.10
          Chromosome Location:
          Chr.1: 207752038 - 207795516 on Build GRCh38
          UniGene ID:
          Hs.510402
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533866

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD46
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002389.4 NP_002380.3 5 772
          NM_153826.3 NP_722548.1 5 772
          NM_172350.2 NP_758860.1 5 772
          NM_172351.2 NP_758861.1 5 772
          NM_172352.2 NP_758862.1 5 772
          NM_172353.2 NP_758863.1 5 772
          NM_172359.2 NP_758869.1 5 772
          NM_172361.2 NP_758871.1 5 772
          XM_011509563.1 XP_011507865.1 5 772
          XM_011509564.1 XP_011507866.1 5 772
          XM_017001308.1 XP_016856797.1 5 772
          XM_017001309.1 XP_016856798.1 5 772
          XM_017001310.1 XP_016856799.1 5 772
          GenBank mRNA A18585.1 - 5 630
          AK222822.1 - 5 698
          AK291227.1 - 5 726
          BC030594.2 - 5 712 4823040
          BX537451.1 - 4 2268
          BX640613.1 - 5 754
          BX649050.1 - 5 743
          D84105.1 - 5 618
          EF076055.1 - 5 630
          EF076056.1 - 5 630
          EF076057.1 - 5 630
          EF076058.1 - 5 630
          M58050.1 - 5 631
          S51940.1 - 5 684
          X59405.1 - 5 742
          X59406.1 - 5 701
          X59407.1 - 5 701
          X59408.1 - 5 742
          X59409.1 - 5 701
          X59410.1 - 5 701
          Y00651.1 - 5 640

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell mediated immunity
          proteolysis
          complement activation, classical pathway
          single fertilization
          regulation of Notch signaling pathway
          positive regulation of gene expression
          negative regulation of gene expression
          regulation of complement activation
          interleukin-10 production
          positive regulation of interleukin-10 production
          sequestering of extracellular ligand from receptor
          positive regulation of T cell proliferation
          negative regulation of catalytic activity
          positive regulation of memory T cell differentiation
          innate immune response
          positive regulation of regulatory T cell differentiation
          negative regulation of complement activation
          viral entry into host cell
          positive regulation of transforming growth factor beta production
          virus receptor activity
          complement binding
          endopeptidase activity
          enzyme inhibitor activity
          receptor activity
          protein binding
          cadherin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0